493171
O-Methylisourea-13C hydrochloride
99 atom % 13C
Synonym(s):
Carbamimidic, methyl ester, monohydrochloride-13C acid
About This Item
isotopic purity
99 atom % 13C
Quality Level
form
solid
technique(s)
protein expression: suitable
mp
122-132 °C (lit.)
mass shift
M+1
SMILES string
Cl.CO[13C](N)=N
InChI
1S/C2H6N2O.ClH/c1-5-2(3)4;/h1H3,(H3,3,4);1H/i2+1;
InChI key
MUDVUWOLBJRUGF-CGOMOMTCSA-N
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service