Skip to Content
Merck
All Photos(1)

Documents

488801

Sigma-Aldrich

Diethylamine-15N hydrochloride

98 atom % 15N

Synonym(s):

N-ethyl-Ethanamine-15N hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)215NH · HCl
CAS Number:
Molecular Weight:
110.59
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

Quality Level

form

solid

mp

227-230 °C (lit.)

mass shift

M+1

SMILES string

Cl.CC[15NH]CC

InChI

1S/C4H11N.ClH/c1-3-5-4-2;/h5H,3-4H2,1-2H3;1H/i5+1;

InChI key

HDITUCONWLWUJR-LJJZSEGWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service