Skip to Content
Merck
All Photos(1)

Key Documents

486604

Sigma-Aldrich

Mevalonolactone-2-13C

99 atom % 13C, 98% (CP)

Synonym(s):

DL-Mevalonic-2-13C lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H10O3
CAS Number:
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.473 (lit.)

bp

150 °C/5 mmHg (lit.)

mp

28 °C (lit.)

mass shift

M+1

SMILES string

CC1(O)CCOC(=O)[13CH2]1

InChI

1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i4+1

InChI key

JYVXNLLUYHCIIH-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell P Swift et al.
PLoS pathogens, 16(2), e1008316-e1008316 (2020-02-15)
Malaria parasites rely on a plastid organelle for survival during the blood stages of infection. However, the entire organelle is dispensable as long as the isoprenoid precursor, isopentenyl pyrophosphate (IPP), is supplemented in the culture medium. We engineered parasites to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service