Skip to Content
Merck
All Photos(1)

Key Documents

486183

Sigma-Aldrich

Pyridine-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H515N
CAS Number:
Molecular Weight:
80.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor pressure

10 mmHg ( 13.2 °C)

isotopic purity

98 atom % 15N

Assay

99% (CP)

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.990 g/mL at 25 °C

mass shift

M+1

SMILES string

c1cc[15n]cc1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i6+1

InChI key

JUJWROOIHBZHMG-PTQBSOBMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Danila A Barskiy et al.
The journal of physical chemistry letters, 9(11), 2721-2724 (2018-05-10)
Hyperpolarization techniques based on the use of para-hydrogen provide orders of magnitude signal enhancement for magnetic resonance spectroscopy and imaging. The main drawback limiting widespread applicability of para-hydrogen-based techniques in biomedicine is the presence of organometallic compounds (the polarization transfer
Yiling Chen et al.
Environmental science & technology, 50(1), 249-258 (2015-12-05)
A combined experimental-theoretical approach was taken to elucidate the reduction mechanisms of five representative aromatic N-oxides (ANOs) by Fe(II)-tiron complex and to identify the rate-limiting step. Based on the possible types of complexes formed with the reductant, three groups of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service