Skip to Content
Merck
All Photos(1)

Key Documents

485705

Sigma-Aldrich

Formic-d acid

95 wt. % in H2O, 98 atom % D

Synonym(s):

Deuterated formic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCO2H
CAS Number:
Molecular Weight:
47.03
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

concentration

95 wt. % in H2O

refractive index

n20/D 1.369

bp

100.8 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.246 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]C(O)=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)/i1D

InChI key

BDAGIHXWWSANSR-MICDWDOJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Celia Muñoz-Muñoz et al.
Journal of analytical toxicology, 44(1), 36-40 (2019-07-03)
An amphetamine interference was observed during the development of an liquid chromatography-high-resolution mass spectrometry (LC-HRMS) multi-class confirmation method for the determination of 47 drugs and metabolites in urine. The interference passed all qualitative criteria for amphetamine leading to potential false-positive
Masayuki Iguchi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(67), 17017-17021 (2017-08-05)
Kinetic studies on the catalytic reaction mechanism of formic acid (FA) dehydrogenation were performed in the presence of a water-soluble iridium complex bearing a 4,4'-dihydroxy-2,2'-bipyridine ligand. Determination of kinetic isotope effects revealed that a shift of the rate-limiting step at
Ying Zhang et al.
The Analyst, 145(13), 4442-4451 (2020-06-13)
Hydrotreatment is extensively used for the production of clean fuel. Attaining an understanding of the structural conversion of the nitrogen species during hydrotreatment is very challenging due to the compositional complexity and the absence of a proper characterization method. In
Agnieszka Piegat et al.
Molecules (Basel, Switzerland), 24(17) (2019-08-25)
The structure of acylated chitosan derivatives strongly determines the properties of obtained products, influencing their hydrodynamic properties and thereby their solubility or self-assembly susceptibility. In the present work, the significance of slight changes in acylation conditions on the structure and
Rose M Gathungu et al.
Analytical chemistry, 88(18), 9103-9110 (2016-08-18)
Lipids from different classes sometimes can exhibit the same exact mass upon electrospray ionization; this presents an analytical challenge in lipidomics. In the negative ionization mode, for example, this can occur with phosphatidylcholines (PCs) and phosphatidylserines (PSs), making them indistinguishable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service