Skip to Content
Merck
All Photos(2)

Key Documents

482552

Sigma-Aldrich

Polypyrrole

greener alternative

doped, 5 wt % dispersion in H2O, conductivity >0.005 S/cm (dried cast film)

Synonym(s):

PPy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

contains

proprietary organic acids as dopant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5 wt % dispersion in H2O

greener alternative category

SMILES string

[nH]1cccc1

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

General description

Polypyrrole (PPy) is a conductive and highly stable polymer, doped with proprietary organic acids. It may be prepared by standard electrochemical technique.[1] PPy may also be prepared by reacting β-napthalene sulfonic acid (NSA) and ammonium peroxo-disulphate in aqueous medium.[2] The charges on the surfaces can be easily modified by doping the polymer during its synthesis.[1]Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid has been reported.[3]
Polypyrrole(PPy) is a π-conjugated conductive polymer that can be electrochemically synthesized and deposited on a variety of surfaces. The deposited PPy can be used in a wide range of sensor based applications due to the following properties[4]:
  • Ion exchange capacity.
  • Electrochromic effects.
  • Redox activity.
  • Corrosion resistant.
  • Catalytic activity.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product enables flexible high-performance energy conversion and storage and thus has been enhanced for energy efficiency. Click here for more information.

Application

Conducting polymers may be used in electronics, chemical sensors.[5]
PPy can be used in the development of devices including:
  • conductive sensors for quality monitoring[6]
  • blended with nanotubes for supercapacitors[7]
  • nanowire actuators[8]
  • biosensors for drug delivery[9]
  • CO2 absorption materials[7]

Packaging

Packaged in poly bottles

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical sensors based on conducting polymer?polypyrrole.
Ramanavicius A, et al.
Electrochimica Acta, 51(27), 6025-6037 (2006)
Polypyrrole nanowire actuators.
Berdichevsky Y and Lo Y
Advanced Materials, 18(1), 122-125 (2006)
Conducting polymers in electronic chemical sensors
Janata J and Josowicz M
Nature Materials, 2, 19-24 (2003)
Biosensing and drug delivery by polypyrrole.
Geetha S, et al.
Analytica Chimica Acta, 568(1-2), 119-125 (2006)
N-doped polypyrrole-based porous carbons for CO2 capture.
Sevilla M, et al.
Advances in Functional Materials, 21(14), 2781-2787 (2011)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

See All

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Specification sheet for CAS 30604-81-0 lists film conductivity. There is no info regarding processing parameter (anneal temp, time) or film characteristics (thickness, roughness). Please provide more info on how film conductivity measurements were done.

    1 answer
    1. Unfortunately, the data regarding the processing parameter or film characteristics is not available for this product. Testing performed on this product is found on the Product Specification Sheet. Please see the document below:
      https://www.sigmaaldrich.com/specification-sheets/339/964/482552-BULK_______ALDRICH__.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service