Skip to Content
Merck
All Photos(3)

Documents

476870

Sigma-Aldrich

α-Cyano-4-hydroxycinnamic acid

99%

Synonym(s):

alpha-Cyano-4-hydroxycinnamic acid, α-CCA, α-CHCA, α-Cyano, 4-HCCA, ACCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=C(CN)CO2H
CAS Number:
Molecular Weight:
189.17
Beilstein:
3271427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

245-250 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)\C(=C\c1ccc(O)cc1)C#N

InChI

1S/C10H7NO3/c11-6-8(10(13)14)5-7-1-3-9(12)4-2-7/h1-5,12H,(H,13,14)/b8-5+

InChI key

AFVLVVWMAFSXCK-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Cyano-4-hydroxycinnamic acid is the commonly used matrix for analyzing peptides by matrix-assisted laser desorption/ionization mass spectrometry. It inhibits the monophenolase activity and diphenolase activity of mushroom tyrosinase.

Application

α-Cyano-4-hydroxycinnamic acid (CHC) was used in encapsulation of CHC into NaY zeolite. It was used as matrix to investigate the activity of the paclitaxel derivatives using several well-established in vitro angiogenesis assays.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusaku Hioki et al.
Journal of mass spectrometry : JMS, 48(11), 1217-1223 (2013-11-22)
We describe here an optimization study of the sample preparation conditions for sensitive detection of peptides by matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS). Among many factors in the conditions, we varied the percent acetonitrile in the peptide solution, the percent
Encapsulation of α-cyano-4-hydroxycinnamic acid into a NaY zeolite.
Vilaca N, et al.
J. Mater. Sci., 46(23), 7511-7516 (2011)
Claudia Ryppa et al.
International journal of pharmaceutics, 368(1-2), 89-97 (2008-11-11)
The alpha(v)beta(3) integrin is overexpressed on proliferating endothelial cells such as those present in growing tumors as well as on tumor cells of various origins. Tumor-induced angiogenesis can be inhibited in vivo by antagonizing the alpha(v)beta(3) integrin with small peptides
Inhibitory effects of α-cyano-4-hydroxycinnamic acid on the activity of mushroom tyrosinase.
Qiu L, et al.
Food Chemistry, 112(3), 609-613 (2009)
Mohammadreza Shariatgorji et al.
Analytical chemistry, 84(16), 7152-7157 (2012-08-07)
D(4)-α-Cyano-4-hydroxycinnamic acid (D(4)-CHCA) has been synthesized for use as a matrix for matrix-assisted laser desorption ionization-mass spectrometry (MALDI-MS) and MALDI-MS imaging (MSI) of small molecule drugs and endogenous compounds. MALDI-MS analysis of small molecules has historically been hindered by interference

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service