Recommended Products
Assay
90%
refractive index
n20/D 1.432 (lit.)
bp
248 °C (lit.)
density
1.141 g/mL at 25 °C (lit.)
functional group
carboxylic acid
ester
SMILES string
CCOC(=O)CCC(O)=O
InChI
1S/C6H10O4/c1-2-10-6(9)4-3-5(7)8/h2-4H2,1H3,(H,7,8)
InChI key
LOLKAJARZKDJTD-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Applied microbiology and biotechnology, 101(15), 5973-5984 (2017-05-14)
Succinic acid (SA) was esterified with ethanol using Candida antarctica lipase B immobilized on acrylic resin at 40 and 50 °C. Enzyme activity in the reaction medium was assured prior to reaction experiments. Reaction-equilibrium experiments were performed for varying initial molalities
Foods (Basel, Switzerland), 10(2) (2021-02-13)
Brandy de Jerez is a European Geographical Indication for grape-derived spirits aged in oak casks that have previously contained any kind of Sherry wine and, therefore, are known as Sherry Casks®. Wood compounds have a substantial influence in the quality
Characterization of different fruit wines made from cacao, cupuassu, gabiroba, jaboticaba and umbu.
LWT--Food Science and Technology, 43(10), 1564-1572 (2010)
Journal of agricultural and food chemistry, 65(35), 7736-7748 (2017-08-02)
Cognac wine spirit has a complex composition in volatile compounds which contributes to its organoleptic profile. This work focused on the batch distillation process and, in particular, on volatile compounds specifically produced by chemical reactions during the distillation of Cognac
Nature genetics, 50(8), 1086-1092 (2018-07-18)
The hereditary cancer syndromes hereditary leiomyomatosis and renal cell cancer (HLRCC) and succinate dehydrogenase-related hereditary paraganglioma and pheochromocytoma (SDH PGL/PCC) are linked to germline loss-of-function mutations in genes encoding the Krebs cycle enzymes fumarate hydratase and succinate dehydrogenase, thus leading
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service