Skip to Content
Merck
All Photos(2)

Key Documents

468347

Sigma-Aldrich

(R)-(+)-2-Bromo-3-methylbutyric acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(Br)CO2H
CAS Number:
Molecular Weight:
181.03
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

optical activity

[α]22/D +21°, c = 37 in benzene

bp

90-100 °C/0.5 mmHg (lit.)

mp

35-40 °C (lit.)

SMILES string

CC(C)[C@@H](Br)C(O)=O

InChI

1S/C5H9BrO2/c1-3(2)4(6)5(7)8/h3-4H,1-2H3,(H,7,8)/t4-/m1/s1

InChI key

UEBARDWJXBGYEJ-SCSAIBSYSA-N

Application

(R)-(+)-2-Bromo-3-methylbutyric acid can be used:
  • As an anionic counterpart of morpholinium based chiral ionic liquids applicable in the Heck arylation reaction of 2,3-dihydrofuran.
  • As a starting material in the synthesis of fructosyl peptide oxidase inhibitors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of chiral ionic liquids on palladium-catalyzed Heck arylation of 2, 3-dihydrofuran
Roszak R, et al.
Applied Catalysis A: General, 409, 148-155 (2011)
Synthesis and inhibitory activity of substrate-analog fructosyl peptide oxidase inhibitors
Watanabe B, et al.
Bioorganic & Medicinal Chemistry Letters, 25(18), 3910-3913 (2015)
J M Te Koppele et al.
The Biochemical journal, 252(1), 137-142 (1988-05-15)
The stereoselectivity of purified rat GSH transferases towards alpha-bromoisovaleric acid (BI) and its amide derivative alpha-bromoisovalerylurea (BIU) was investigated. GSH transferase 2-2 was the only enzyme to catalyse the conjugation of BI and was selective for the (S)-enantiomer. The conjugation
M Polhuijs et al.
Biochemical pharmacology, 38(22), 3957-3962 (1989-11-15)
The glutathione (GSH) conjugation of (R)-and (S)-alpha-bromoisovaleric acid (BI) in the rat in vivo, and its stereoselectivity, have been characterized. After administration of racemic [1-14C]BI two radioactive metabolites were found in bile: only one of the possible diastereomeric BI-GSH conjugates
M Polhuijs et al.
Biochemical pharmacology, 44(7), 1249-1253 (1992-10-06)
Glutathione (GSH) conjugation of the separate enantiomers of five 2-bromocarboxylic acids and some of their urea derivatives by rat liver GSH transferases (GSTs) was studied. The liver cytosolic fraction conjugated all compounds, except for (R)-2-bromoisovaleric acid, with a variable degree

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service