All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C4H6N4
CAS Number:
Molecular Weight:
110.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
143-147 °C (lit.)
SMILES string
Nc1ccnc(N)n1
InChI
1S/C4H6N4/c5-3-1-2-7-4(6)8-3/h1-2H,(H4,5,6,7,8)
InChI key
YAAWASYJIRZXSZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wenbo Zhou et al.
European journal of medicinal chemistry, 96, 269-280 (2015-04-23)
Therapeutics of metastatic or triple-negative breast cancer are still challenging in clinical. Herein we demonstrated the design and optimization of a series of hybrid of 2,4-diaminopyrimidine and arylthiazole derivatives for their anti-proliferative properties against two breast cancer cell lines (MCF-7
Hydrogen-bonded supramolecular structures in co-crystals of ?-or ?-diketone enols with 2, 6-diaminopyridine or 2, 4-diaminopyrimidine.
Bertolasi V, et al.
New. J. Chem., 26(11), 1559-1566 (2002)
Wilhelm Maximilian Hützler et al.
Acta crystallographica. Section C, Structural chemistry, 71(Pt 3), 229-238 (2015-03-04)
The results of seven cocrystallization experiments of the antithyroid drug 6-methyl-2-thiouracil (MTU), C(5)H(6)N(2)OS, with 2,4-diaminopyrimidine, 2,4,6-triaminopyrimidine and 6-amino-3H-isocytosine (viz. 2,6-diamino-3H-pyrimidin-4-one) are reported. MTU features an ADA (A = acceptor and D = donor) hydrogen-bonding site, while the three coformers show
Eugen F Mesaros et al.
Bioorganic & medicinal chemistry letters, 21(1), 463-466 (2010-11-16)
The synthesis and biological evaluation of potent and selective anaplastic lymphoma kinase (ALK) inhibitors from a novel class of 2,4-diaminopyrimidines, incorporating 2,3,4,5-tetrahydro-benzo[d]azepine fragments, is described. An orally bioavailable analogue (18) that displayed antitumor efficacy in ALCL xenograft models in mice
Craig A Zificsak et al.
Bioorganic & medicinal chemistry letters, 21(13), 3877-3880 (2011-06-03)
The incorporation of R,R-1,2-diaminocyclohexane at C4 in a series of 2,4-diaminopyrimidines led to a number of ALK inhibitors in which optimized activity was achieved by conversion of the 2-amino group into a methanesulfonamide. Tumor growth inhibition was observed when an
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service