Skip to Content
Merck
All Photos(1)

Key Documents

466409

Sigma-Aldrich

Poly(ethylene glycol) sorbitol hexaoleate

Synonym(s):

Polyethoxylated sorbitol hexaoleate, Polyoxyethylene sorbitan hexaoleate, Sorbeth hexaoleate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

refractive index

n20/D 1.471 (lit.)

viscosity

200 cP(25 °C)(lit.)

hydroxyl value

35‑55 mg KOH/g

saponification value

80‑90 mg KOH/g

solubility

propylene glycol: insoluble

density

1.03 g/mL at 25 °C (lit.)

HLB

10

SMILES string

O([C@@H]([C@H](OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H](OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)COCCOC(=O)CCCCCCC\C=C/CCCCCCCC)[C@@H](OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)COCCOC(=O)CCCCCCC\C=C/CCCCCCCC)CCOC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C126H230O18/c1-7-13-19-25-31-37-43-49-55-61-67-73-79-85-91-97-119(127)137-105-103-133-115-117(135-107-109-139-121(129)99-93-87-81-75-69-63-57-51-45-39-33-27-21-15-9-3)125(143-113-111-141-123(131)101-95-89-83-77-71-65-59-53-47-41-35-29-23-17-11-5)126(144-114-112-142-124(132)102-96-90-84-78-72-66-60-54-48-42-36-30-24-18-12-6)118(136-108-110-140-122(130)100-94-88-82-76-70-64-58-52-46-40-34-28-22-16-10-4)116-134-104-106-138-120(128)98-92-86-80-74-68-62-56-50-44-38-32-26-20-14-8-2/h49-60,117-118,125-126H,7-48,61-116H2,1-6H3/b55-49-,56-50-,57-51-,58-52-,59-53-,60-54-/t117-,118+,125-,126-/m1/s1

InChI key

LAIORVRDCREVSA-AMAZOXQHSA-N

Application

Surfactant.
Emulsifier for fats and oils.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service