Skip to Content
Merck
All Photos(3)

Documents

464171

Sigma-Aldrich

Zirconium(IV) hydroxide

97%

Synonym(s):

R Zirconium Hydroxide, Zirconic acid, Zirconium hydroxide, Zirconium tetrahydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OH)4
CAS Number:
Molecular Weight:
159.25
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

density

3.25 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O[Zr](O)(O)O

InChI

1S/4H2O.Zr/h4*1H2;/q;;;;+4/p-4

InChI key

HAIMOVORXAUUQK-UHFFFAOYSA-J

General description

Zirconium(IV) hydroxide is a white to off-white solid that is sparinglysoluble in water. It has relatively high specific surface area, ion exchange capacity, good thermalstability, and dielectric properties.

Application

Zirconium(IV) hydroxide can be used as:
  • A starting material to prepare sulfated zirconia catalyst for liquid-phase dehydration of sorbitol.
  • To fabricate coated magnetic materials for the removal of anionic dyes from solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and Efficient Removal of Anionic Dyes from Solution by Zirconium(IV) Hydroxide-Coated Magnetic Materials
Yanyan Hu and Runping Han
Journal of Chemical and Engineering Data, 64, 791-799 (2019)
Liquid-phase dehydration of sorbitol to isosorbide using sulfated zirconia as a solid acid catalyst
Nazmul Abedin Khan, et al
Applied Catalysis A: General, 452, 34-38 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service