Skip to Content
Merck
All Photos(1)

Key Documents

463469

Sigma-Aldrich

2-Amino-3,5-dibromobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H2(Br)2CHO
CAS Number:
Molecular Weight:
278.93
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

130-135 °C (lit.)

functional group

aldehyde
bromo

SMILES string

Nc1c(Br)cc(Br)cc1C=O

InChI

1S/C7H5Br2NO/c8-5-1-4(3-11)7(10)6(9)2-5/h1-3H,10H2

InChI key

RCPAZWISSAVDEA-UHFFFAOYSA-N

General description

2-Amino-3,5-dibromobenzaldehyde has been identified as one of the oxidation product of bromhexine by controlled potential electrolysis followed by HPLC-UV and GC-MS. It participates in the Friedländer condensation of C-β−glycosylic ketones to form 2-substituted quinoline derivatives.

Application

2-Amino-3,5-dibromobenzaldehyde may be employed in the preparation of tetradentate Schiff base ligands, via condensation with aliphatic diamines. These ligands forms nickel (II) and oxovanadium(IV) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Turchán et al.
Talanta, 73(5), 913-919 (2007-10-31)
A complete electrochemical study and a novel electroanalytical procedure for bromhexine quantitation are described. Bromhexine in methanol/0.1molL(-1) Britton-Robinson buffer solution (2.5/97.5) shows an anodic response on glassy carbon electrode between pH 2 and 7.5. By DPV and CV, both peak
Subbiah Nagarajan et al.
Carbohydrate research, 345(14), 1988-1997 (2010-08-07)
Regioselective facile one-pot synthesis of 16 different sugar-based quinoline, naphthyridine, and xanthone derivatives is reported. The compounds are characterized by NMR spectroscopy and elemental analysis. The beta-Anomeric form of the sugar moiety was identified from (1)H NMR studies. Antimicrobial studies
Khosro Mohammadi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 146, 221-227 (2015-03-31)
The tetradentate Schiff base ligands (L(1)-L(4)), were synthesized by reaction between 2-amino-3,5-dibromobenzaldehyde and aliphatic diamines. Then, nickel and oxovanadium(IV) complexes of these ligands were synthesized and characterized by (1)H NMR, Mass, IR, UV-Vis spectroscopy and thermogravimetry. The kinetic parameters of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service