Skip to Content
Merck
All Photos(1)

Key Documents

46188

Sigma-Aldrich

Farnesal, mixture of isomers

technical

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrienal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein:
1723427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

impurities

≤3% water

refractive index

n20/D >1.4920 (lit.)
n20/D 1.497

bp

126-129 °C/3.5 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)\C=C(/C)CC\C=C(/C)CC\C=C(/C)C

InChI

1S/C15H24O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11-12H,5-6,8,10H2,1-4H3/b14-9+,15-11+

InChI key

YHRUHBBTQZKMEX-YFVJMOTDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Farnesal has been reported as specific lipid substrate for aldo-keto reductase 1B10 (AKR1B10). The incubation of farnesol with the protoplast of Botryococcus braunii B race strain leads to the formation of farnesal and 3-hydroxy-2,3-dihydrofarnesal.

Application

Farnesal was used in the synthesis of sesquiterpene nanaimoal.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoyo Sakata et al.
Experimental & applied acarology, 29(3-4), 279-291 (2003-11-26)
The composition of oil gland exudates from two oribatid mites, Trhypochthoniellus sp. and Trhypochthonius japonicus, was studied with reference to the related species Trhypochthoniellus crassus. Trhypochthoniellus sp. contained a mixture of seven compounds; (Z,Z)-6,9-heptadecadiene, geranial, 3-hydroxybenzene-1,2-dicarbaldehyde (gamma-acaridial), neryl formate, neral
Kenji Shimomura et al.
Journal of chemical ecology, 34(4), 467-477 (2008-03-21)
The southern cowpea weevil, Callosobruchus chinensis (Coleoptera: Bruchidae), is a major pest of stored legumes in warm temperate and tropical climates. The female sex attractant pheromone was extracted from filter-paper shelters taken from containers that housed virgin females. The extracts
H Inoue et al.
Biochemical and biophysical research communications, 196(3), 1401-1405 (1993-11-15)
Farnesal and 3-hydroxy-2,3-dihydrofarnesal (3-hydroxy-3,7,11-trimethyl-6,10-dodecadiene-1-al) were formed from farnesol when the alcohol was incubated with the protoplast of Botryococcus braunii B race strain. This fact suggests the existence of farnesal hydratase in the alga. Feeding experiments showed that both farnesal and
W R Tschantz et al.
The Journal of biological chemistry, 276(4), 2321-2324 (2000-11-18)
Prenylated proteins contain either a 15-carbon farnesyl or a 20-carbon geranylgeranyl isoprenoid covalently attached via a thioether bond to a cysteine residue at or near their C terminus. As prenylated proteins comprise up to 2% of the total protein in
Kenji Shimomura et al.
Journal of chemical ecology, 36(8), 824-833 (2010-07-08)
Callosobruchus rhodesianus (Pic) (Coleoptera: Chrysomelidae: Bruchinae) is a pest of stored legumes through the Afro-tropical region. In laboratory bioassays, males of C. rhodesianus were attracted to volatiles collected from virgin females. Collections were purified by various chromatographic techniques, and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service