Skip to Content
Merck
All Photos(1)

Key Documents

460346

Sigma-Aldrich

Strontium

dendritic pieces, purified by distillation, 99.9% trace metals basis

Synonym(s):

Strontium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr
CAS Number:
Molecular Weight:
87.62
EC Number:
MDL number:
UNSPSC Code:
12141504
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9% trace metals basis

form

dendritic pieces

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

23 μΩ-cm, 20°C

bp

1384 °C (lit.)

mp

757 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

[Sr]

InChI

1S/Sr

InChI key

CIOAGBVUUVVLOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dendritic strontium is a form of strontium metal that has a unique, tree-like or branching morphology. It is often made by electrodeposition. Dendritic strontium is a silvery-white metal that is highly reactive with water and oxygen, and it readily tarnishes in air. It has several important physical properties, including a melting point of 769 °C and a density of 2.54 g/cm3.

Application

Dendritic strontium has a number of important uses including use as a component of alloys, as a material for pyrotechnic devices, as a reagent in the production of ferrite magnets, and as a dopant in the production of semiconductors.
Dendritic-strontium can be applied in the solid state synthesis of a Zintl phase, SrSn3Sb4, with an anionic channel framework of 30-membered rings.[1]It can also be used to make strontium-potassium-niobate (Sr2KNb5O15, SKN) which is of great interest for pyroelectric sensors and various electro-optic devices.[2]
Used to make Strontium-potassium-niobate (Sr2KNb5O15, SKN). SKN is of great interest for pyroelectric sensors and various electro-optic devices.2

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Sr 2 KNb 5 O 15 Thin Films by Chemical Solution Deposition Method.
Sakamoto W, et al.
Journal of Materials Research, 14(04), 1495-1502 (1999)
Emily Y Tsui et al.
Nature chemistry, 5(4), 293-299 (2013-03-21)
Redox-inactive metals are found in biological and heterogeneous water oxidation catalysts, but, at present, their roles in catalysis are not well understood. Here, we report a series of high-oxidation-state tetranuclear-dioxido clusters comprising three manganese centres and a redox-inactive metal (M).
Zuzana Saidak et al.
Pharmacology & therapeutics, 136(2), 216-226 (2012-07-24)
Osteoporosis is an important age-related bone disease characterized by increased bone turnover with insufficient bone formation relative to bone resorption. According to the current understanding of this disorder, anti-resorptive and anabolic drugs have been developed for therapeutic intervention. Another therapeutic
Wenjie Zhang et al.
Biomaterials, 34(13), 3184-3195 (2013-02-06)
Both surface chemistry and topography have significant influence on good and fast osseointegration of biomedical implants; the main goals in orthopeadic, dental and maxillofacial surgeries. A surface modification strategy encompassing the use of bioactive trace elements together with surface micron/nano-topographical
Strontium and bone.
W E Cabrera et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 14(5), 661-668 (1999-05-13)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service