Skip to Content
Merck
All Photos(1)

Key Documents

456179

Sigma-Aldrich

Phenethyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NCO
CAS Number:
Molecular Weight:
147.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.522 (lit.)

bp

210 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

functional group

amine
isocyanate
phenyl

SMILES string

O=C=NCCc1ccccc1

InChI

1S/C9H9NO/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2

InChI key

HACRKYQRZABURO-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silica gel high-performance liquid chromatography for the simultaneous determination of propranolol and 4-hydroxypropranolol enantiomers after chiral derivatization.
M J Wilson et al.
Journal of chromatography, 310(2), 424-430 (1984-10-12)
J Gal et al.
Drug metabolism and disposition: the biological fate of chemicals, 9(6), 557-560 (1981-11-01)
Chiral secondary alcohols were treated with (S)-(-)-1-phenylethyl isocyanate. For each racemic alcohol, the resulting diastereomeric urethane derivatives were resolved on flexible fused-silica capillary GLC columns with retention times of 15 min or less. Derivatization of individual enantiomers showed that the
A new synthesis of isocyanates.
Gittos MW, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2, 141-143 (1976)
Peter Rose et al.
World journal of gastroenterology, 11(26), 3990-3997 (2005-07-05)
Hydrogen sulfide (H(2)S) is a prominent gaseous constituent of the gastrointestinal (GI) tract with known cytotoxic properties. Endogenous concentrations of H(2)S are reported to range between 0.2-3.4 mmol/L in the GI tract of mice and humans. Considering such high levels
H Spahn-Langguth et al.
Journal of analytical toxicology, 15(6), 327-331 (1991-11-01)
For the enantiospecific assay of propranolol in biological material, formation of diastereomeric derivatives is one possible approach. The aim of the present study was the development and optimization of three analytical methods based on different chiral reagents: phenylethylisocyanate and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service