Skip to Content
Merck
All Photos(1)

Key Documents

451045

Sigma-Aldrich

Iodine

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Diiodine, Molecular iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

Assay

99.999% trace metals basis

form

beads

resistivity

1.3E15 μΩ-cm

particle size

−10 mesh

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodine is an efficient, inexpensive, non-toxic, easy-to-handle electrophilic reagent used in various organic transformations. The electrophilic properties of iodine are effectively utilized in the cyclization of heteroatom-containing alkenyl and alkynyl derivatives. It is also used as an oxidizing agent in the aromatization of α,β-unsaturated cyclohexenone derivatives, and their heterocyclic analogs.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Molecular Iodine-Mediated Cyclization of Tethered Heteroatom-Containing Alkenyl or Alkynyl Systems
Malose Jack Mphahlele
Molecules (Basel), 14 (2009)
Molecular Iodine?An Expedient Reagent for Oxidative Aromatization Reactions of ?,?-Unsaturated Cyclic Compounds
Malose Jack Mphahlele
Molecules (Basel), 14 (2009)
Tim I M Korevaar et al.
The Journal of clinical endocrinology and metabolism, 98(11), 4382-4390 (2013-09-17)
Premature delivery is an important risk factor for child mortality and psychiatric, metabolic, and cardiovascular disease later in life. In the majority of cases, the cause of prematurity cannot be identified. Currently, it remains controversial whether abnormal maternal thyroid function
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and
Michael B Zimmermann
Thyroid : official journal of the American Thyroid Association, 17(9), 829-835 (2007-10-25)
Iodine is required for the production of thyroid hormones, which are essential for normal brain development, and the fetus, newborn, and young child are particularly vulnerable to iodine deficiency. The iodine requirement increases during pregnancy and recommended intakes are in

Articles

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service