Skip to Content
Merck
All Photos(1)

Key Documents

450383

Sigma-Aldrich

Polystyrene

average Mw ~400,000 by GPC

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~400,000 by GPC

mp

212 °C

SMILES string

c1(ccccc1)C=C

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Applications of Polystyrene in Industrial Chemistry: This comprehensive review discusses the synthesis, characteristics, and wide-ranging applications of polystyrene, emphasizing its role in academic research and industrial usage (T Arfin et al., 2015).
  • Upcycling Polystyrene: This article explores innovative approaches to recycling polystyrene, reducing environmental impact and advancing sustainability in material sciences (JC Capricho et al., 2022).
  • Potential Toxicity of Polystyrene Microplastic Particles: Examines the potential impacts of polystyrene microplastics on human health at the cellular level, important for public health and safety (J Hwang et al., 2020).

Physical form

90% isotactic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Megan Farrell et al.
Colloids and surfaces. B, Biointerfaces, 81(2), 468-475 (2010-08-31)
Dextran and polyethylene glycol (PEG) are often covalently bound to the surface of polydimethylsiloxane (PDMS) for the purpose of modifying its hydrophilicity and biocompatibility. In this work, the effects of the dextran and PEG on the morphology, wetting, and surface
Keiji Hirota et al.
Colloids and surfaces. B, Biointerfaces, 87(2), 293-298 (2011-06-28)
Our previous results on the phagocytic activity of alveolar macrophages (Mϕs) toward poly(lactic-co-glycolic) acid microspheres (PLGA MS) loaded with the anti-tuberculosis agent rifampicin (R-PLGA MS) suggest that the phagocytosis of R-PLGA MS enhances the phagocytic activity of Mϕ cells. To
M Jönsson-Niedziółka et al.
Lab on a chip, 11(3), 490-496 (2010-11-26)
Environmental air monitoring is of great interest due to the large number of people concerned and exposed to different possible risks. From the most common particles in our environment (e.g. by-products of combustion or pollens) to more specific and dangerous
Robert W Mann
Journal of forensic sciences, 54(6), 1229-1230 (2009-09-08)
An inexpensive and effective method for articulating a dry human spine is described. By constructing a Styrofoam spine tray, analysts can now accurately position and align each vertebra in correct anatomical order, allow for gaps because of missing vertebrae, and
Chongyang Shen et al.
Journal of contaminant hydrology, 134-135, 1-11 (2012-05-12)
This study examined the detachments of nano- and micro-sized colloids from primary minima in the presence of cation exchange by laboratory column experiments. Colloids were initially deposited in columns packed with glass beads at 0.2 M CaCl(2) in the primary

Articles

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

The present publication focuses on synthesis and characterization of nanothick, chemically grafted polymer films (polymer brushes) on various inorganic and polymeric substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service