Skip to Content
Merck
All Photos(1)

Key Documents

447439

Sigma-Aldrich

1-Methyl-L-tryptophan

95%, for peptide synthesis

Synonym(s):

(S)-2-Amino-3-(1-methyl-1H-indol-3-yl)propanoic acid, L-(-)-1-Methyltryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

1-Methyl-L-tryptophan, 95%

Quality Level

Assay

95%

form

solid

optical activity

[α]24/D −8.0°, c = 2 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

242-245 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cn1cc(C[C@H](N)C(O)=O)c2ccccc12

InChI

1S/C12H14N2O2/c1-14-7-8(6-10(13)12(15)16)9-4-2-3-5-11(9)14/h2-5,7,10H,6,13H2,1H3,(H,15,16)/t10-/m0/s1

InChI key

ZADWXFSZEAPBJS-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methyl-L-tryptophan can be used:
  • In the preparation of L-tryptophan-based ligand for regioselective copper catalyzed N2-arylation of 1,2,3-triazoles.
  • As a key starting material for the total synthesis of (−)-ardeemin.
  • In the synthesis of cyclic peptide-capped gold nanoparticles as drug delivery systems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a L?Tryptophan?Based Ligand for Regioselective Copper Catalyzed N2?Arylation of 1, 2, 3?Triazoles.
Lopes A B, et al.
ChemistrySelect, 2(22), 6544-6548 (2017)
Stefanie Siegert et al.
PloS one, 6(11), e27618-e27618 (2011-11-24)
Adaptive immune responses are initiated when T cells encounter antigen on dendritic cells (DC) in T zones of secondary lymphoid organs. T zones contain a 3-dimensional scaffold of fibroblastic reticular cells (FRC) but currently it is unclear how FRC influence
Total synthesis of (?)-ardeemin.
He B, et al.
The Journal of Organic Chemistry, 74(1), 298-304 (2008)
Cyclic peptide-capped gold nanoparticles as drug delivery systems.
Nasrolahi Shirazi A, et al.
Molecular Pharmaceutics, 10(2), 500-511 (2012)
Juliana Croitoru-Lamoury et al.
PloS one, 6(2), e14698-e14698 (2011-03-02)
The kynurenine pathway (KP) of tryptophan metabolism is linked to antimicrobial activity and modulation of immune responses but its role in stem cell biology is unknown. We show that human and mouse mesenchymal and neural stem cells (MSCs and NSCs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service