Skip to Content
Merck
All Photos(2)

Key Documents

442305

Sigma-Aldrich

Bis(ethylenediamine)copper(II) hydroxide solution

1.0 M in H2O

Synonym(s):

Copper(II)-ethylenediame complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(H2NCH2CH2NH2)2(OH)2
CAS Number:
Molecular Weight:
217.76
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

reagent type: catalyst
core: copper

concentration

1.0 M in H2O

density

1.086 g/mL at 25 °C

SMILES string

O[Cu]O.NCCN.NCCN

InChI

1S/2C2H8N2.Cu.2H2O/c2*3-1-2-4;;;/h2*1-4H2;;2*1H2/q;;+2;;/p-2

InChI key

WATCRQGYOIZIHC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Bis(ethylenediamine)copper(II) hydroxide solution is a stable complex formed by copper (Cu) ions with ethylenediamine. It is a transition metal solution that can be used as a catalyst for electrochemical reduction process.

Application

Bis(ethylenediamine)copper(II) hydroxide has been used to evaluate cation exchange capacity (CEC) of clays by copper absorption.
Copper(II) ethylenediamine complex may be used to evaluate the cation exchange capacity (CEC) of clay materials. It may also be used to modify polyoxoniobate, which is a class of polyoxometalates (POMs) that finds potential application in photocatalytic H2 production.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CEC of clays: measurement by adsorption of a copper ethylenediamine complex
Bergaya F and Vayer M
Applied Clay Science, 12(3), 275-280 (1997)
Determination of the cation exchange capacity (CEC) of clay minerals using the complexes of copper (II) ion with triethylenetetramine and tetraethylenepentamine.
Lorenz P, et al.
Clays and Clay Minerals, 47, 386-388 (1999)
A copper (ii)-ethylenediamine modified polyoxoniobate with photocatalytic H2 evolution activity under visible light irradiation.
Wang Z, et al.
Dalton Transactions, 41(33), 9882-9884 (2012)
Far-infrared and electronic spectra of some bis (ethylenediamine) and related complexes of copper (II) and the relevance of these data to tetragonal distortion and bond strengths.
Lever ABP and Mantovani E
Inorganic Chemistry, 10(4), 817-826 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service