Skip to Content
Merck
All Photos(2)

Key Documents

441783

Sigma-Aldrich

Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate)

98%

Synonym(s):

Antioxidant 1010, Irganox 1010, Pentaerythritol tetrakis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H2[C(CH3)3]2CH2CH2CO2CH2]4C
CAS Number:
Molecular Weight:
1177.63
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

115-118 °C (dec.) (lit.)

SMILES string

CC(C)(C)c1cc(CCC(=O)OCC(COC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)(COC(=O)CCc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)COC(=O)CCc4cc(c(O)c(c4)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

InChI

1S/C73H108O12/c1-65(2,3)49-33-45(34-50(61(49)78)66(4,5)6)25-29-57(74)82-41-73(42-83-58(75)30-26-46-35-51(67(7,8)9)62(79)52(36-46)68(10,11)12,43-84-59(76)31-27-47-37-53(69(13,14)15)63(80)54(38-47)70(16,17)18)44-85-60(77)32-28-48-39-55(71(19,20)21)64(81)56(40-48)72(22,23)24/h33-40,78-81H,25-32,41-44H2,1-24H3

InChI key

BGYHLZZASRKEJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentaerythritol tetrakis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate) (PT) is a sterically hindered phenolic antioxidant and a heat stabilizer mainly used to enhance the oxidative resistance and protection against the toxicity of solvents. It has a glass transition temperature of 50 0C and can be annealed at room temperature.

Application

PT can be used as an additive to protect biaxially-oriented propylene (BOPP) capacitor film from thermal oxidation on UV exposure. It can also be used as a standard material to study the gas-cluster ion sputtering (GCIS) using X-ray photoelectron microscopy (XPS) and Atomic force microscopy (AFM).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of UV treatment on the dielectric strength of BOPP capacitor film
Conference Record of the 2006 IEEE International Symposium on Electrical Insulation, 36(5), 314-317 (2006)
Effect of carbon nanotubes on the global aging behavior of beta-nucleated polypropylene random copolymers for absorbers of solar-thermal collectors.
Wallner GM, et al.
Solar Energy (2018)
Upcycling of polypropylene?The influence of polyethylene impurities.
Kamleitner F, et al.
Polym. Eng. Sci., 57(12), 1374-1381 (2017)
Gas-cluster ion sputtering: Effect on organic layer morphology.
Goodwin CM, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 36(5), 051507-051507 (2018)
W Limm et al.
Food additives and contaminants, 12(4), 609-624 (1995-07-01)
The effect of mixing on the migration of Irganox 1010 antioxidant from polypropylene and high-density polyethylene to water and corn oil was compared at 77, 100, and 135 degrees C. Irganox 1010 migration to water is enhanced almost five-fold by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service