Skip to Content
Merck
All Photos(2)

Key Documents

441643

Sigma-Aldrich

3-Fluorophenylboronic acid

≥95.0%

Synonym(s):

(3-Fluorophenyl-1-yl)boronic acid, 3-Fluorobenzeneboronic acid, m-Fluorophenylboronic acid, m-fluoro-Benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4B(OH)2
CAS Number:
Molecular Weight:
139.92
Beilstein:
3030632
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

214-218 °C (lit.)

functional group

fluoro

SMILES string

OB(O)c1cccc(F)c1

InChI

1S/C6H6BFO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4,9-10H

InChI key

KNXQDJCZSVHEIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recently used to make novel liquid crystalline fluorobiphenylcyclohexenes and difluoroterphenyls by palladium-catalyzed cross-couplings also used in the synthesis of o-phenylphenols as potent leukotriene B4 receptor agonists.

Other Notes

Contains varying amounts of anhydride

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

o-phenylphenols: potent and orally active leukotriene B4 receptor antagonists.
M J Sofia et al.
Journal of medicinal chemistry, 36(24), 3978-3981 (1993-11-26)
Liq. Cryst., 18, 1-1 (1995)
Journal of Materials Chemistry, 5, 2239-2239 (1995)
Chihiro Takei et al.
Chemical & pharmaceutical bulletin, 66(4), 368-374 (2018-04-03)
Previous studies have shown that reversible chemical bond formation between phenylboronic acid (PBA) and 1,3-diol can be utilized as the driving force for the preparation of layer-by-layer (LbL) films. The LbL films composed of a PBA-appended polymer and poly(vinyl alcohol)
Mikhail Y Vorona et al.
Materials (Basel, Switzerland), 13(8) (2020-04-26)
Anthracene-based semiconductors have attracted great interest due to their molecular planarity, ambient and thermal stability, tunable frontier molecular orbitals and strong intermolecular interactions that can lead to good device field-effect transistor performance. In this study, we report the synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service