Skip to Content
Merck
All Photos(1)

Key Documents

432997

Sigma-Aldrich

Poly(dimethylsiloxane), hydroxy terminated

viscosity 18,000-22,000 cSt 

Synonym(s):

PDMS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

Quality Level

refractive index

n20/D 1.404

viscosity

18,000-22,000 cSt

density

0.97 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(O)O

Looking for similar products? Visit Product Comparison Guide

Application

Component of silicone room temperature vulcanizing system that incorporates reinforcing aids or fillers.

Features and Benefits

Can be polymerized using small amounts of multi-functional silanes, aided by organometallic salts.

Physical form

Dihydroxy functional reactive silicone fluid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service