Skip to Content
Merck
All Photos(1)

Key Documents

432040

Sigma-Aldrich

Benzyloxyacetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CO2H
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

137-139 °C/0.6 mmHg (lit.)

density

1.162 g/mL at 25 °C (lit.)

SMILES string

OC(=O)COCc1ccccc1

InChI

1S/C9H10O3/c10-9(11)7-12-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)

InChI key

GRZHHTYDZVRPIC-UHFFFAOYSA-N

General description

In vitro activities of phenoxy and benzyloxyacetic acid derivatives as antisickling agents has been investigated by the application of quantitative structure activity relationship (QSAR) of Hansch-type.

Application

Benzyloxyacetic acid may be used for the following syntheses:
  • mixed benzyloxyacetic pivalic anhydride
  • chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterification
  • tetraaqua(1,10-phenanthroline-[κ]2N,N′)magnesium(II) bis[(2,4-dichlorophenyl)acetate]
  • as ligand in the preparation of diaquabis(benzyloxyacetato)copper(II) complex

Other Notes

may contain chloroacetic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Mahran
Bollettino chimico farmaceutico, 139(2), 73-80 (2000-08-02)
Quantitative structure activity relationship of Hansch-type has been applied to develop correlation between the calculated physicochemical properties and the in vitro activities of phenoxy and benzyloxyacetic acid derivatives as antisickling agents. The antisickling effect of these compounds was first reported
An inexpensive carbohydrate derivative used as a chiral auxiliary in the synthesis of a-hydroxy carboxylic acids.
Yu H, et al.
Tetrahedron, 58(38), 7663-7669 (2002)
A highly effective asymmetric synthesis of a-hydroxy acids by alkylation of chiral n-(benzyloxyacetyl)-trans-2, 5-bis (methoxymethoxymethyl) pyrrolidine.
Enomoto M, et al.
Tetrahedron Letters, 26(10), 1343-1344 (1985)
Diaquabis (benzyloxyacetato) copper (II).
Sun S-L, et al.
Acta Crystallographica Section E, Structure Reports Online, 64(5), 1052-1052 (2008)
Xiao-Min Hao et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 8), m1052-m1052 (2008-01-01)
In the mononuclear title complex, [Mg(C(12)H(8)N(2))(H(2)O)(4)](C(8)H(5)Cl(2)O(2))(2), each Mg(II) ion is hexa-coordinated by two N atoms from a 1,10-phenanthroline ligand [Mg-N = 2.233 (2) Å] and four water mol-ecules [Mg-OW = 2.033 (2) and 2.043 (1) Å] in a distorted octa-hedral geometry. A twofold rotation axis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service