About This Item
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.409 (lit.)
bp
140 °C (lit.)
density
0.726 g/mL at 25 °C (lit.)
SMILES string
CCCCC(C)C(C)C
InChI
1S/C9H20/c1-5-6-7-9(4)8(2)3/h8-9H,5-7H2,1-4H3
InChI key
WBRFDUJXCLCKPX-UHFFFAOYSA-N
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
195.8 °F - closed cup
Flash Point(C)
91 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service