Skip to Content
Merck
All Photos(1)

Key Documents

421669

Sigma-Aldrich

Asp-Phe

96%, for peptide synthesis

Synonym(s):

L-Aspartyl-L-phenylalanine, Aspartame acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)CO2H
CAS Number:
Molecular Weight:
280.28
Beilstein:
2817528
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

Asp-Phe, 96%

Quality Level

Assay

96%

form

powder

optical activity

[α]20/D +12.5°, c = 1 in 0.5 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

236-239 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C13H16N2O5/c14-9(7-11(16)17)12(18)15-10(13(19)20)6-8-4-2-1-3-5-8/h1-5,9-10H,6-7,14H2,(H,15,18)(H,16,17)(H,19,20)/t9-,10-/m0/s1

InChI key

YZQCXOFQZKCETR-UWVGGRQHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Lipton et al.
Metabolism: clinical and experimental, 40(12), 1337-1345 (1991-12-01)
The dipeptide sweetener aspartame (N-L-alpha-aspartyl-L-phenylalanine, 1-methyl ester; alpha-APM) is relatively stable in dry powder form. However, when exposed to elevated temperature, extremes of pH and/or moisture, alpha-APM is converted into a variety of products. In aqueous solution alpha-APM decomposes to
Montserrat Andújar-Sánchez et al.
FEBS letters, 581(18), 3449-3454 (2007-07-10)
Somatic angiotensin I-converting enzyme (s-ACE) plays a central role in blood pressure regulation and has been the target of most antihypertensive drugs. A displacement isothermal titration calorimetry method has been used to accurately determine the binding constant of three strong
Y In et al.
Chemical & pharmaceutical bulletin, 48(3), 374-381 (2000-03-22)
As part of the series investigating the structural features of C-terminal amidated amino acids and peptides, three crystal structures of Z-Gly-Phe-NH2, Tyr-Lys-NH2, and Asp-Phe-NH2 were analyzed by the X-ray diffraction method, and their molecular conformations and intermolecular interactions were investigated.
Kimber van Vliet et al.
Nutrients, 12(6) (2020-07-01)
Phenylketonuria and tyrosinemia type 1 are treated with dietary phenylalanine (Phe) restriction. Aspartame is a Phe-containing synthetic sweetener used in many products, including many 'regular' soft drinks. Its amount is (often) not declared; therefore, patients are advised not to consume
D Grobelny et al.
Biochemical and biophysical research communications, 128(2), 960-964 (1985-04-30)
Aspartame (L-aspartyl-L-phenylalanine methyl ester, is a widely used artificIal sweetener. In humans and other animals aspartame is initially hydrolyzed to L-aspartyl-L-phenylalanine by intestinal esterases. L-Aspartyl-L-phenylalanine inhibits angiotensin converting enzyme purified from rabbit lungs with a Ki of 11 +/- 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service