Skip to Content
Merck
All Photos(1)

Key Documents

417041

Sigma-Aldrich

S-Alpine-Borane®

97%

Synonym(s):

B-Isopinocampheyl-9-borabicyclo[3.3.1]nonane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31B
CAS Number:
Molecular Weight:
258.25
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

97%

optical activity

[α]22/D +20°, c = 12 in THF

bp

>55 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1[C@H](CC2CC1C2(C)C)B3C4CCCC3CCC4

InChI

1S/C18H31B/c1-12-16-10-13(18(16,2)3)11-17(12)19-14-6-4-7-15(19)9-5-8-14/h12-17H,4-11H2,1-3H3/t12-,13+,14-,15+,16-,17-/m0/s1

InChI key

VCDGSBJCRYTLNU-NEXGVSGLSA-N

General description

S-Alpine-Borane® is a chiral reducing agent, synthesized from (-)-α-pinene via hydroboration.

Application

S-Alpine-Borane® may be used in the preparation of (-)-(4S)-5-methyl-2-hexyn-4-ol, an intermediate for the synthesis of (+)-Prelog-Djerassi lactonic aldehyde.

Legal Information

Alpine-Borane is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Liq. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of [2,3] sigmatropic (Wittig) rearrangements in synthesis. The synthesis of (+)-Prelog-Djerassi lactone.
Tsai DJS, and Midland MM.
Journal of the American Chemical Society, 107(13), 3915-3918 (1985)
Matteson DS
Stereodirected Synthesis with Organoboranes, 346-347 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service