Skip to Content
Merck
All Photos(2)

Key Documents

416835

Sigma-Aldrich

4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N · HCl
CAS Number:
Molecular Weight:
195.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

solid

mp

202-203.5 °C (lit.)

functional group

phenyl

SMILES string

Cl[H].C1CC(=CCN1)c2ccccc2

InChI

1S/C11H13N.ClH/c1-2-4-10(5-3-1)11-6-8-12-9-7-11;/h1-6,12H,7-9H2;1H

InChI key

POGWXTJNUCZEPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenyl-1,2,3,6-tetrahydropyridine fragment containing benzamide derivatives have been prepared. Presence of the 4-phenyl-1,2,3,6-tetrahydropyridine fragment improves the inhibitory potential of benzamide analogs against poly(ADP-ribose) polymerase-1 (PARP-1).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyota Fujita et al.
PloS one, 4(9), e7247-e7247 (2009-10-01)
It has been shown that molecular hydrogen (H(2)) acts as a therapeutic antioxidant and suppresses brain injury by buffering the effects of oxidative stress. Chronic oxidative stress causes neurodegenerative diseases such as Parkinson's disease (PD). Here, we show that drinking
Jin Gyu Choi et al.
Journal of ethnopharmacology, 134(2), 414-421 (2011-01-05)
Polygalae radix, the root of Polygala tenuifolia Willd, has commonly been used for the treatment of amnesia and anxiety in traditional Korean medicine. The aim of this study was to investigate its neuroprotective effects and possible mechanisms of action in
Jennifer M King et al.
Life sciences, 89(17-18), 638-643 (2011-08-30)
l-3,4-Dihydroxyphenylalanine (L-DOPA) is the most effective symptomatic treatment for Parkinson's disease (PD), but PD patients usually experience a successful response to L-DOPA therapy followed by a progressive loss of response. L-DOPA efficacy relies on its decarboxylation by aromatic l-amino acid
C A Dodd et al.
Toxicology and industrial health, 25(1), 25-39 (2009-03-26)
The pyrethroid insecticide permethrin and the organophosphate insecticide chlorpyrifos can experimentally produce Parkinson's disease (PD)-associated changes in the dopaminergic nigrostriatal pathway, short of frank degeneration, although at doses considerably higher than from a likely environmental exposure. The ability of permethrin
Tultul Nayyar et al.
The European journal of neuroscience, 30(2), 207-216 (2009-08-08)
Parkinson's Disease (PD) is marked by prominent motor symptoms that reflect striatal dopamine insufficiency. However, non-motor symptoms, including depression, are common in PD. It has been suggested that these changes reflect pathological involvement of non-dopaminergic systems. We examined regional changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service