Skip to Content
Merck
All Photos(1)

Key Documents

414506

Sigma-Aldrich

2-(Tributylstannyl)furan

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30OSn
CAS Number:
Molecular Weight:
357.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

90 °C/0.1 mmHg (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccco1

InChI

1S/C4H3O.3C4H9.Sn/c1-2-4-5-3-1;3*1-3-4-2;/h1-3H;3*1,3-4H2,2H3;

InChI key

SANWDQJIWZEKOD-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Nowakowska-Oleksy et al.
Journal of fluorescence, 21(1), 169-178 (2010-07-14)
A few new phenoxazine-based conjugated monomers were synthesized, characterized, and successfully used as semiconducting materials. The phenoxazine-based oligomers have low ionization potentials or high-lying HOMO levels (~4.7 eV), which were estimated from cyclic voltammetry. Conjugated oligomers offer good film-forming, mechanical and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service