Skip to Content
Merck
All Photos(1)

Documents

408360

Sigma-Aldrich

Di(trimethylolpropane) tetraacrylate

average Mw 466

Synonym(s):

Bistrimethylolpropane tetraacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(H2C=CHCO2CH2)2C(C2H5)CH2]2O
CAS Number:
Molecular Weight:
466.52
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mw 466

contains

1000 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.479 (lit.)

density

1.101 g/mL at 25 °C (lit.)

SMILES string

CCC(COCC(CC)(COC(=O)C=C)COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

InChI

1S/C24H34O9/c1-7-19(25)30-15-23(11-5,16-31-20(26)8-2)13-29-14-24(12-6,17-32-21(27)9-3)18-33-22(28)10-4/h7-10H,1-4,11-18H2,5-6H3

InChI key

XRMBQHTWUBGQDN-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Weng et al.
JCI insight, 5(20) (2020-09-11)
Focal adhesion kinase (FAK) is an important mediator of extracellular matrix-integrin mechano-signal transduction that regulates cell motility, survival, and proliferation. As such, FAK is being investigated as a potential therapeutic target for malignant and fibrotic diseases, and numerous clinical trials
A Horrer et al.
Nanoscale, 9(44), 17378-17386 (2017-11-03)
We report on a compact and cost-effective integrated label-free biosensor configuration which is based on the refractive index sensitivity of the localized surface plasmon resonance (LSPR) of gold nanostructures. Aiming for compactification and miniaturization of the sensor, arrays of nanodiscs
Yekaterina A Miroshnikova et al.
Nature cell biology, 18(12), 1336-1345 (2016-11-08)
Increased overall survival for patients with glioma brain tumours is associated with mutations in the metabolic regulator isocitrate dehydrogenase 1 (IDH1). Gliomas develop within a mechanically challenged microenvironment that is characterized by a dense extracellular matrix (ECM) that compromises vascular
Dennis Graf et al.
Materials (Basel, Switzerland), 13(11) (2020-06-11)
3D inkjet printing is moving from a technology of rapid prototyping to rapid manufacturing. The introduction of ultraviolet curable composites filled with functional ceramics could expand the possibilities of this technology. In this work, a simple and scalable process was
Jonathon M Muncie et al.
Developmental cell, 55(6), 679-694 (2020-11-19)
Embryogenesis is directed by morphogens that induce differentiation within a defined tissue geometry. Tissue organization is mediated by cell-cell and cell-extracellular matrix (ECM) adhesions and is modulated by cell tension and tissue-level forces. Whether cell tension regulates development by modifying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service