Skip to Content
Merck
All Photos(1)

Documents

406023

Sigma-Aldrich

Diethylzinc solution

1.0 M in heptane

Synonym(s):

Zincdiethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2Zn
CAS Number:
Molecular Weight:
123.51
Beilstein:
3587207
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in heptane

bp

98 °C

density

0.74 g/mL at 25 °C

SMILES string

CC[Zn]CC

InChI

1S/2C2H5.Zn/c2*1-2;/h2*1H2,2H3;

InChI key

HQWPLXHWEZZGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylzinc solution can be used in the synthesis of:
  • Bis(pyridylpyrrolyl)zinc luminescent complexes.
  • A versatile building block, 5-(ketoaryl)thiazole.
  • ZnxCd1-xSe nanocrystals having high luminescence properties.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition-Tunable ZnxCd1-xSe nanocrystals with high luminescence and stability.
Xinhua Z, et al.
Journal of the American Chemical Society, 125(28), 8589-8594 (2003)
Pyridylpyrrolides as alternatives to cyclometalated phenylpyridine ligands: synthesis and characterization of luminescent zinc and boron pyridylpyrrolide complexes.
Klappa JJ, et al.
Dalton Transactions, 883-891 (2004)
Practical synthesis of a highly functionalized thiazole ketone.
Frey LF, et al.
Tetrahedron, 59(33), 6363-6373 (2003)
Yan Yin et al.
The Journal of organic chemistry, 72(15), 5731-5736 (2007-06-22)
Intramolecular hydroamination of alkynyl amides was effected by a catalytic amount of Et2Zn (20 mol %) to form indole derivatives, and a tandem cyclization/nucleophilic addition procedure involving reaction of the indole zinc salt intermediate with acid chlorides or halides was
Su-Xi Wang et al.
Chemical & pharmaceutical bulletin, 55(7), 1011-1013 (2007-07-03)
Newly synthesized polymer-supported chiral amino alcohol catalysts 5a and 5b have been proved to be effective for the enantioselective addition of diethylzinc to aldehydes, affording the corresponding sec-alcohols in moderate enantiomeric excesses with good to excellent yields. The recycled catalyst

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service