Skip to Content
Merck
All Photos(1)

Key Documents

404330

Sigma-Aldrich

1-Ethynyl-4-fluorobenzene

99%

Synonym(s):

(4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne, (p-Fluorophenyl)acetylene, (p-Fluorophenyl)ethyne, 1-Fluoro-4-ethynylbenzene, 2-(4-Fluorophenyl)acetylene, 4-Ethynylfluorobenzene, 4-Fluoroethynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4C≡CH
CAS Number:
Molecular Weight:
120.12
Beilstein:
1099285
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

refractive index

n20/D 1.516 (lit.)

bp

55-56 °C/40 mmHg (lit.)

mp

26-27 °C (lit.)

density

1.048 g/mL at 25 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C#C

InChI

1S/C8H5F/c1-2-7-3-5-8(9)6-4-7/h1,3-6H

InChI key

QXSWHQGIEKUBAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethynyl-4-fluorobenzene (pFAB) is a fluorobenzene derivative. Trianionic pincer alkylidyne complex catalyzed polymerization of 1-ethynyl-4-fluorobenzene has been reported. The surface of the iodophenyl-terminated organic monolayers has been modified by Sonogashira coupling of the iodophenyl groups found on the surface with pFAB. Dimerization catalyzed by Y[N(TMS)2]3 and 4-chloroaniline gave exclusive formation of one of three possible enediynes in excellent yield.

Application

1-Ethynyl-4-fluorobenzene may be used in the synthesis of the following:
  • 3-(4-Fluorophenyl)-1H-isochromen-1-one by reacting with 2-iodobenzoic acid in the presence of copper catalyst.
  • 4-Bromo-3,5-difluoro-2,6-bis-(4-fluoro-phenylethynyl)-pyridine by reacting with 2,4,6-tribromo-3,5-difluoropyridine by palladium catalyzed Sonogashira reaction.
  • 1-(4-Fluorophenyl)-3-methoxy-3-(4-methoxyphenyl)propan-1-one by reacting with p-anisaldehyde.
  • 5-(4-Fluorophenyl)-1-(2-nitrobenzyl)-1H-1,2,3-triazole by reacting with 1-(bromomethyl)-2-nitrobenzene in the presence of sodium azide.
1-Ethynyl-4-fluorobenzene, a substituted phenylacetylene, may be used in the synthesis of aryl acetylenes, via Sonogashira type cross coupling reaction with various haloarenes in the presence of bis(μ-iodo)bis((-)-sparteine)dicopper(I) catalyst.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyhalogenoheterocyclic compounds. Part 53. Sonogashira reactions of 2, 4, 6-tribromo-3, 5-difluoropyridine.
Howardb JAK.
ARKIVOC (Gainesville, FL, United States), 11, 46-55 (2007)
Soumya Sarkar et al.
Journal of the American Chemical Society, 134(10), 4509-4512 (2012-02-23)
Synthesis, characterization, and catalytic alkyne polymerization results for the first trianionic pincer alkylidyne complex, [(t)BuOCO]W≡CC(CH(3))(3)(THF)(2) (6), are described. Complex 6 is a highly active catalyst for the polymerization of acetylenes and exhibits a high turnover number (4371), activity (1.05 ×
Novel regio-and stereoselective dimerization of terminal alkynes catalyzed by rare-earth silylamide.
Komeyama K, et al.
Synthesis, 7, 1062-1066 (2004)
Pd-catalyzed coupling reaction on the organic monolayer: Sonogashira reaction on the silicon (111) surfaces.
Qu M, et al.
Applied Surface Science, 255(5), 2608-2612 (2008)
Bis (μ-iodo) bis ((-)-sparteine) dicopper (I) catalyzed Sonogashira-type reaction under palladium and phosphine-free conditions.
Priyadarshini S, et al.
Tetrahedron Letters, 52(14), 1615-1618 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service