Skip to Content
Merck
All Photos(1)

Key Documents

393207

Sigma-Aldrich

Diethyl 3,4-pyrroledicarboxylate

98%

Synonym(s):

3,4-Bis(ethoxycarbonyl)pyrrole, 3,4-Diethyl 1H-pyrrole-3,4-dicarboxylate, Diethyl 1H-pyrrole-3,4-dicarboxylate, Diethyl pyrrole-3,4-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO4
CAS Number:
Molecular Weight:
211.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

147-149 °C (dec.) (lit.)

functional group

ester

SMILES string

CCOC(=O)c1c[nH]cc1C(=O)OCC

InChI

1S/C10H13NO4/c1-3-14-9(12)7-5-11-6-8(7)10(13)15-4-2/h5-6,11H,3-4H2,1-2H3

InChI key

QKXBVVVYILIRDO-UHFFFAOYSA-N

General description

Diethyl 3,4-pyrroledicarboxylate (Diethyl 1H-pyrrole-3,4-dicarboxylate) is a 3,4-pyrrole dicarboxylic diester. It has been prepared by the hydrolysis of diethyl 1-benzoyl-3,4-pyrroledicarboxylate and confirmed by IR, NMR and mass spectra.

Application

Diethyl 3,4-pyrroledicarboxylate may be used in the synthesis of trisubstituted pyrroles like diethyl 1-hydroxymethyl-3,4-pyrroledicarboxylate and diethyl 1-benzoyloxyrmethyl-3,4-pyrroledicarboxylate. It is also used to prepare pyrrole copolymer soft actuators with reduced electrochemical creep and actuating strain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Pyrrole Copolymer Soft Actuators Prepared by Electrochemical Polymerization Pyrrole and Diethyl 3, 4-pyrroledicarboxylate.
Ogihara S, et al.
Mol. Cryst. Liq. Cryst., 566(1), 158-164 (2012)
Helena K Kim et al.
Journal of psychiatric research, 76, 59-65 (2016-02-20)
Chronic N-methyl-d-aspartate (NMDA) administration to rats may be a model to investigate excitotoxicity mediated by glutamatergic hyperactivity, and lithium has been reported to be neuroprotective. We hypothesized that glutamatergic hyperactivity in chronic NMDA injected rats would cause mitochondrial dysfunction and
Manabu Taneike et al.
PloS one, 11(3), e0152628-e0152628 (2016-03-31)
Mammalian target of rapamycin complex 1 (mTORC1) is a key regulator of cell growth, proliferation and metabolism. mTORC1 regulates protein synthesis positively and autophagy negatively. Autophagy is a major system to manage bulk degradation and recycling of cytoplasmic components and
Pyrrole Chemistry. XV. The Chemistry of Some 3, 4-Disubstituted Pyrroles.
Groves JK, et al.
Canadian Journal of Chemistry, 51(7), 1089-1098 (1973)
Mustafa I Ahmed et al.
The Journal of thoracic and cardiovascular surgery, 152(4), 1059-1070 (2016-07-29)
Recent studies have demonstrated improved outcomes in patients receiving early surgery for degenerative mitral regurgitation (MR) rather than adhering to conventional guidelines for surgical intervention. However, studies providing a mechanistic basis for these findings are limited. Left ventricular (LV) myocardium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service