All Photos(1)
About This Item
Linear Formula:
CH3OC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
197.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
125-130 °C (lit.)
functional group
carboxylic acid
nitro
SMILES string
COc1ccc(c(c1)C(O)=O)[N+]([O-])=O
InChI
1S/C8H7NO5/c1-14-5-2-3-7(9(12)13)6(4-5)8(10)11/h2-4H,1H3,(H,10,11)
InChI key
URADKXVAIGMTEG-UHFFFAOYSA-N
Related Categories
General description
5-Methoxy-2-nitrobenzoic acid is a substituted 2-nitrobenzoic acid. It has been reported to be one of the microbial degradation metabolite of cypermethrin, a synthetic pyrethroid pesticide.
Application
5-Methoxy-2-nitrobenzoic acid may be used as starting material in the synthesis of the following:
- dictyoquinazol A, a neuroprotective compound
- 5-methoxyantranilic acid by catalytic reduction
- pyrrolobenzodiazepines
5-Methoxy-2-nitrobenzoic acid may be used in the synthesis of the following N-phenylethyl-2-nitrobenzamides:
- N-phenylethyl-5-methoxy-2-nitrobenzamide
- N-(4-methoxyphenylethyl)-5-methoxy-2-nitrobenzamide
- N-(3,4-dimethoxyphenylethyl)-5-methoxy-2-nitrobenzamide
- N-(phenethyl)-5-chloro-2-nitrobenzamide
- N-(3,4-dimethoxyphenethyl)-5-chloro-2-nitrobenzamide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Total synthesis of neuroprotective dictyoquinazol A, B, and C.
Oh CH and Song CH.
Synthetic Communications, 37(19), 3311-3317 (2007)
Yin LB, et al.
Biodegradation of Hazardous and Special Products (Intech), 1-16 (2013)
A Novel Synthesis of 5, 6-Dihydroindazolo [3,2-a] isoquinolines and Their Relative Compounds via Tin (II) Chloride Dihydrate as Reducing Agent.
Kuo CY and Wu MJ.
J. Chin. Chem. Soc., 52(5), 965-974 (2005)
Synthesis of pyrrolobenzodiazepines via the PIFA oxidation of amines. Synthesis of 8-deoxy DC-81.
Kraus GA and Melekhov A.
Tetrahedron, 54(39), 11749-11754 (1998)
Derivatives of 6-methoxy-2-mercaptoquinazole-4-one as compounds with potential anti-tubercularactivity.
Murav'eva KM, et al.
Pharmaceutical Chemistry Journal, 1(8), 456-458 (1967)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service