Skip to Content
Merck
All Photos(1)

Key Documents

391433

Sigma-Aldrich

tert-Butylamine

≥99.5%

Synonym(s):

2-Amino-2-methylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

vapor pressure

5.7 psi ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

716 °F

expl. lim.

9.8 %

refractive index

n20/D 1.377 (lit.)

bp

46 °C (lit.)

mp

−67 °C (lit.)

solubility

water: miscible 1000 g/L at 25 °C
alcohol: miscible(lit.)
chloroform: soluble(lit.)
organic solvents: soluble(lit.)

density

0.696 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)(C)N

InChI

1S/C4H11N/c1-4(2,3)5/h5H2,1-3H3

InChI key

YBRBMKDOPFTVDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butylamine (TBA) is an aliphatic primary amine. Its enthalpy of combustion and formation has been studied. Its gas phase acidity and gas phase basicity has been evaluated. The ozonation of tert-butylamine has been reported. Its influence on the aqueous beta-cyclodextrin (β-CD): pyrene complex has been examined by steady-state fluorescence measurements.

Application

tert-Butylamine (t-butylamine) may be used in the synthesis of Zn2-SnO4 nanoparticles. It may be used to terminate the polymerization reaction during the synthesis of the following polymers (DAB =1,4-diaminobutane, BA=Boronic acid , PAA=functional poly(amido amine), ABOL=4-aminobutanol, Bn=Benzyl):
  • (DAB-BA-PAA)
  • (ABOL-BA-PAA)
  • (DAB-Bn-PAA)
tert-Butylamine (TBA) is suitable for use as an indicator molecule in the study of NMR-based pH determination method, free of glass electrode errors in highly basic media. It is suitable for use in the synthesis of methyl β-ketophosphonate, titanium dioxide nanoparticles (TiO2 NPs) and samarium oxide (Sm2O3) nanospheres and nanorods. It may be used in the following studies:
  • As liquid hydrocarbon promoter in the systematic study on methane hydrate formation and dissociation.
  • As an example in the study of the effect of amines in influencing atmospheric H2SO4-H2O nucleation.
  • Hydroamination of terminal alkynes.
  • To measure the surface acidity of solid catalysts by temperature-programmed desorption.
  • As a reductive nitrogen source in the atomic layer deposition technique used to deposit TaN thin films from TaCl5 and TaBr5.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-36.4 °F - closed cup

Flash Point(C)

-38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled self-assembly of Sm2O3 nanoparticles into nanorods: simple and large scale synthesis using bulk Sm2O3 powders.
Nguyen TD, et al.
The Journal of Physical Chemistry C, 112(39), 15226-15235 (2008)
Laboratory study on new particle formation from the reaction OH+ SO2: influence of experimental conditions, H2O vapour, NH3 and the amine tert-butylamine on the overall process.
Berndt T, et al.
Atmospheric Chemistry and Physics, 10(15), 7101-7116 (2010)
Gábor Orgován et al.
Journal of pharmaceutical and biomedical analysis, 54(5), 958-964 (2010-12-21)
A set of indicator molecules was selected and applied to elaborate an NMR-based pH determination method, free of glass electrode errors in highly basic media. Accurate measurement of pH values and protonation constants was achieved by a successive build-up of
Enthalpies of combustion and formation of propylamine, isopropylamine, and tert-butylamine.
Smith NK and Good WD
Journal of Chemical and Engineering Data, 12(4), 572-574 (1967)
Christian Balg et al.
Bioorganic & medicinal chemistry, 15(1), 295-304 (2006-10-20)
The aminoacyl-beta-ketophosphonate-adenosines (aa-KPA) are stable analogs of the aminoacyl adenylates, which are high-energy intermediates in the formation of aminoacyl-tRNA catalyzed by aminoacyl-tRNA synthetases (aaRS). We have synthesized glutamyl-beta-ketophosphonate-adenosine (Glu-KPA) and glutaminyl-beta-ketophosphonate-adenosine (Gln-KPA), and have tested them as inhibitors of their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service