Skip to Content
Merck
All Photos(1)

Key Documents

388513

Sigma-Aldrich

5,8-Dihydroxy-1,4-naphthoquinone

technical grade

Synonym(s):

Naphthazarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6O4
CAS Number:
Molecular Weight:
190.15
Beilstein:
880561
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

technical grade

Quality Level

mp

220-230 °C (lit.)

SMILES string

Oc1ccc(O)c2C(=O)C=CC(=O)c12

InChI

1S/C10H6O4/c11-5-1-2-6(12)10-8(14)4-3-7(13)9(5)10/h1-4,11-12H

InChI key

RQNVIKXOOKXAJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Brígida R Pinho et al.
PloS one, 6(8), e24098-e24098 (2011-09-03)
The search of new anti-inflammatory drugs has been a current preoccupation, due to the need of effective drugs, with less adverse reactions than those used nowadays. Several naphthoquinones (plumbagin, naphthazarin, juglone, menadione, diosquinone and 1,4-naphthoquinone), plus p-hydroquinone and p-benzoquinone were
Vishnu K Tandon et al.
Bioorganic & medicinal chemistry letters, 15(13), 3288-3291 (2005-05-26)
A series of (1,4)-naphthoquinono [3,2-c]-1H-pyrazoles and their (1,4)-naphthohydroquinone derivatives 2-7 were synthesized and evaluated for antifungal, antibacterial, and anticancer activities. The structure-activity relationship of these compounds was studied and the results show that the compound 2b exhibited in vitro antifungal
Régis Millet et al.
Journal of medicinal chemistry, 48(22), 7024-7039 (2005-10-28)
The human selenoprotein thioredoxin reductase is involved in antioxidant defense and DNA synthesis. As increased thioredoxin reductase levels are associated with drug sensitivity to cisplatin and drug resistance in tumor cells, this enzyme represents a promising target for the development
An efficient formal synthesis of the human telomerase inhibitor (+/-)-gamma-rubromycin.
Dominea C K Rathwell et al.
Angewandte Chemie (International ed. in English), 48(43), 7996-8000 (2009-08-01)
Piper R Hunt et al.
PloS one, 6(7), e21922-e21922 (2011-07-19)
Hormesis occurs when a low level stress elicits adaptive beneficial responses that protect against subsequent exposure to severe stress. Recent findings suggest that mild oxidative and thermal stress can extend lifespan by hormetic mechanisms. Here we show that the botanical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service