Skip to Content
Merck
All Photos(3)

Key Documents

385980

Sigma-Aldrich

Benzoic anhydride

≥95%

Synonym(s):

Benzoyl anhydride, Benzoyl benzoate, Bis(phenylcarbonyl)ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O
CAS Number:
Molecular Weight:
226.23
Beilstein:
516726
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

mp

38-42 °C (lit.)

density

1.199 g/mL at 25 °C (lit.)

functional group

anhydride
ester
phenyl

SMILES string

O=C(OC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O3/c15-13(11-7-3-1-4-8-11)17-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

CHIHQLCVLOXUJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzoic anhydride can be used as a:
  • Condensation reagent in the synthesis of carboxylic esters from carboxylic acids and alcohols.
  • Coupling reagent in the synthesis of macrolactones.
  • Acylating reagent for acylating sulfonamides, amines, alcohols, and phenols using ZSM-5-SO3H as a catalyst.

It can be also used as a reagent for the preparation of benzyl benzoate , benzylidene dibenzoate , 4-benzoyltoluene .

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZSM-5-SO3H: an efficient catalyst for acylation of sulfonamides amines, alcohols, and phenols under solvent-free conditions
Massah AR, et al.
ISRN Organic Chemistry, 2013(10), 2312-2312 (2013)
TMEDA: efficient and mild catalyst for the acylation of alcohols, phenols and thiols under solvent-free condition
Kadam ST, et al.
Bull. Korean Chem. Soc., 30(5), 1071-1076 (2009)
Formyloxyacetoxyphenylmethane and 1, 1-diacylals as versatile O-formylating and O-acylating reagents for alcohols
Chapman RSL, et al.
Tetrahedron, 74(44), 6442-6452 (2018)
Acylations with long-chain acid anhydrides and acyl chlorides over zeolite beta
Bejblova M, et al.
Topics in Catalysis, 52(1-2), 178-184 (2009)
Isamu Shiina et al.
The Journal of organic chemistry, 69(6), 1822-1830 (2004-04-03)
Various carboxylic esters are obtained at room temperature in excellent yields with high chemoselectivities from nearly equimolar amounts of carboxylic acids and alcohols using 2-methyl-6-nitrobenzoic anhydride with triethylamine by the promotion of a basic catalyst such as 4-(dimethylamino)pyridine. A variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service