Skip to Content
Merck
All Photos(1)

Key Documents

385417

Sigma-Aldrich

3-(Trifluoromethyl)benzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4SO2Cl
CAS Number:
Molecular Weight:
244.62
Beilstein:
2650774
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

88-90 °C/6 mmHg (lit.)

density

1.526 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1cccc(c1)S(Cl)(=O)=O

InChI

1S/C7H4ClF3O2S/c8-14(12,13)6-3-1-2-5(4-6)7(9,10)11/h1-4H

InChI key

ONCAZCNPWWQQMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer supported reagents in synthesis: preparation of bicyclo[2.2.2] octane derivates via Tandem Michael addition reactions and subsequent combinatorial decoration.
S V Ley et al.
Journal of combinatorial chemistry, 2(2), 104-107 (2000-05-16)
Chemistry of aminophenols. Part 2: A general and efficient synthesis of indoles possessing a nitrogen substituent at the C4, C5, C6, and C7 positions.
Dai et al.
Tetrahedron Letters, 43, 7699-7702 (2002)
Preparation of N-(a, ?-unsaturated acyl)-sulfonamides.
Katritzky et al.
ARKIVOC (Gainesville, FL, United States), 4, 115-124 (2009)
Regiocontroled Palladium-Catalysed Direct Arylation at Carbon C2 of Benzofurans using Benzenesulfonyl Chlorides as the Coupling Partners.
Loukotova et al.
ChemCatChem, 5, 1303-1309 (2014)
Synthesis, Characterization and Evaluation of Antibacterial Activity of Six Novel 1, 2, 4-Triazole Derivatives against Standard and Medical Bacteria.
Jacob et al.
Advanced Bacterial Genetics, Cold Spring Harbor, 5(6), 303-318 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service