Skip to Content
Merck
All Photos(2)

Key Documents

363073

Sigma-Aldrich

Poly(vinyl alcohol)

average Mw 31,000-50,000, 87-89% hydrolyzed

Synonym(s):

PVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

crystalline powder
crystals or granules
powder

mol wt

average Mw 31,000-50,000

falling ball

5.2-6.2 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl alcohol) (PVA) is a vinyl based water soluble polymer that can be used as a non-ionic surface active agent. It can also be used as a biodegradable polymer and can be incorporated in adhesives, coatings, textiles ceramics and cosmetics. Its properties include good transparency, chemical resistance, toughness and anti-electrostatic properties.[1][2][3]

Application

PVA can be used in the preparation of microspheres which can be further used as biodegradable microcarriers.[4][5][6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compostable Polymer Properties and Packaging Applications
Plastic Films in Food Packaging, 217-248 (2013)
Gas foamed open porous biodegradable polymeric microspheres
Kim TK, et al.
Biomaterials, 27(2), 152-159 (2006)
Poly (D, L lactic-co-glycolic acid) microspheres as biodegradable microcarriers for pluripotent stem cells
Newman KD and McBurney MW
Biomaterials, 25(26), 5763-5771 (2004)
PLGA/alginate composite microspheres for hydrophilic protein delivery
Zhai P, et al.
Materials Science and Engineering, C, 56(26), 251-259 (2015)
Elisa Rojas-Prats et al.
Pharmaceutics, 13(2) (2021-02-03)
A potent cell division cycle 7 (CDC7) kinase inhibitor, known as PHA-767491, has been described to reduce the transactive response DNA binding protein of 43 KDa (TDP-43) phosphorylation in vitro and in vivo, which is one of the main proteins

Questions

1–2 of 2 Questions  
  1. Once dissolved, does the powder get separated from the solution? If so, how long does it take to be separated from solvent (assuming that solvent is water)?

    1 answer
    1. This product is water soluble up to 20%. The material is expected to stay in solution with proper preparation and storage. General instructions are as follows:
      1. Use unheated water
      2. Add water to vessel and initiate agitation or stirring
      3. Add desired amount of PVA, gradually, up to 20%
      4. Heat solution while stirring to 85°C, hold for 30 minutes

      To maintain the product in solution, do not store at temperatures lower that 15°C.
      Solutions stored at temperatures higher than 24°C should include a biocidal agent.

      Helpful?

  2. Is there information available regarding the solubility of product 363073, which is a PVA (poly vinyl alcohol)? Additionally, how can this particular product be dissolved?

    1 answer
    1. The solubility of product 363073, PVA (poly vinyl alcohol), indicates that it should dissolve in heated water, DMSO, and glycerol. It is expected to be around 40-50mg/mL soluble in hot water at a temperature of approximately 90-95 degrees Celsius.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service