Skip to Content
Merck
All Photos(1)

Key Documents

362026

Sigma-Aldrich

Bis(ethylenedithio)tetrathiafulvalene

98%

Synonym(s):

BEDT-TTF, Bi[5,6-dihydro-1,3-dithiolo[4,5-b][1,4]-dithiine-2-ylidene]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8S8
CAS Number:
Molecular Weight:
384.69
Beilstein:
1653830
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

~260 °C (dec.) (lit.)

SMILES string

C1CSC2=C(S1)S\C(S2)=C3\SC4=C(SCCS4)S3

InChI

1S/C10H8S8/c1-2-12-6-5(11-1)15-9(16-6)10-17-7-8(18-10)14-4-3-13-7/h1-4H2

InChI key

LZJCVNLYDXCIBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(ethylenedithio)tetrathiafulvalene (BEDT-TTF) is an organic superconducting polymer that is used as an electron donor with a superconducting transition temperature (Tc) of 10K.

Application

BEDT-TTF may be used as a counter-anion with tetrathiocyanatocuprate which can be prepared by the galvanostatic electrocrystallization process. It may also be used in the preparation of novel paramagnetic conductors by synthesizing multifunctional molecular materials with manganese containing complexes. BEDT-TTF based doping may be used in the fabrication of a hybrid organic change generation layer which can potentially be used in the development of tandem organic light emitting diodes (OLEDs).
Used as a component for organic superconductors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitaka Kawasugi et al.
Science advances, 5(5), eaav7282-eaav7282 (2019-05-17)
A Mott insulator sometimes induces unconventional superconductivity in its neighbors when doped and/or pressurized. Because the phase diagram should be strongly related to the microscopic mechanism of the superconductivity, it is important to obtain the global phase diagram surrounding the
Mn-Containing Paramagnetic Conductors with Bis (ethylenedithio) tetrathiafulvalene (BEDT-TTF).
Benmansour S, et al.
Magnetochemistry, 3(1), 7-7 (2017)
Anion polarity-induced self-doping in a purely organic paramagnetic conductor, α?-α?-(BEDT-TTF) 2 (PO-CONH-m-C6H4SO3)? H2O where BEDT-TTF is bis (ethylenedithio) tetrathiafulvalene and PO is the radical 2, 2, 5, 5-Tetramethyl-3-pyrrolin-1-oxyl.
Akutsu H, et al.
Polyhedron, 136, 23-29 (2017)
Genta Kawaguchi et al.
Advanced materials (Deerfield Beach, Fla.), 31(2), e1805715-e1805715 (2018-11-09)
Superconducting (SC) devices are attracting renewed attention as the demands for quantum-information processing, meteorology, and sensing become advanced. The SC field-effect transistor (FET) is one of the elements that can control the SC state, but its variety is still limited.
Efficiency enhancement in tandem organic light-emitting devices with a hybrid charge generation layer composed of BEDT-TTF-doped TPBi/mCP/HAT-CN.
Kim DH and Kim TW
Organic Electronics, 15(12), 3452-3457 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service