Skip to Content
Merck
All Photos(1)

Key Documents

360740

Sigma-Aldrich

1,2-Dimethylnaphthalene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
2039376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.615 (lit.)

bp

266-267 °C (lit.)

mp

−2-−1 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc2ccccc2c1C

InChI

1S/C12H12/c1-9-7-8-11-5-3-4-6-12(11)10(9)2/h3-8H,1-2H3

InChI key

QNLZIZAQLLYXTC-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dimethylnaphthalene is a polycyclic aromatic hydrocarbon. Its effect on the naupliar and adult stages of the marine cyclopoid copepod Oithona davisae is reported.

Application

1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secondary organic aerosol formation from photooxidation of naphthalene and alkylnaphthalenes: implications for oxidation of intermediate volatility organic compounds (IVOCs).
Chan AWH, et al.
Atmospheric Chemistry and Physics, 9(9), 3049-3060 (2009)
A Kilanowicz et al.
International journal of occupational medicine and environmental health, 11(4), 305-317 (1999-02-24)
The aim of this study was to investigate the distribution, excretion and metabolism of 1,2-dimethylnaphthalene-[ring-U-3H] in rats. The experiments were performed on 54 male outbred IMP:Wist rats with body weight of 200 g +/- 20%. The compound was given i.p.
Enric Saiz et al.
Environmental pollution (Barking, Essex : 1987), 157(4), 1219-1226 (2009-01-17)
Short-term (24h) exposure experiments have been conducted to determine the effects of two environmental relevant polycyclic aromatic hydrocarbons (PAHs), naphthalene (NAPH) and dimethylnaphthalene (C2-NAPH), on the naupliar and adult stages of the marine cyclopoid copepod Oithona davisae. To resemble more
Nien-Hsin Kao et al.
Marine pollution bulletin, 97(1-2), 319-332 (2015-06-08)
Three fishing harbors were investigated to study the polycyclic aromatic hydrocarbons in the sediments and trace possible anthropogenic sources by identification of cyclic terpenoid biomarkers. Seventeen terpanes, 10 steranes and 10 bicyclic sesquiterpanes in the marine diesel and the three
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service