Skip to Content
Merck
All Photos(1)

Documents

35350

Sigma-Aldrich

1,3-Dichlorobenzene

puriss., ≥99.0% (GC)

Synonym(s):

m-Dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
956618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5 mmHg ( 38.8 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.546 (lit.)
n20/D 1.546

bp

172-173 °C (lit.)

mp

−25-−22 °C (lit.)

density

1.288 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc(Cl)c1

InChI

1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

ZPQOPVIELGIULI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dichlorobenzene is a thermodynamically favoured isomer of 1,4-dichlorobenzene. The heat capacity at constant pressure and density of 1,3-dichlorobenzene was measured in the temperature range from (283.15 to 353.15)K.

Application

1,3-Dichlorobenzene is suitable as carbon and energy supplement for the growth of gram-negative, peritrichously flagellated rod, tentatively identified as an Alcaligenes sp.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A de Bont et al.
Applied and environmental microbiology, 52(4), 677-680 (1986-10-01)
A gram-negative, peritrichously flagellated rod, tentatively identified as an Alcaligenes sp., was isolated from a mixture of soil and water samples by using 1,3-dichlorobenzene as the sole carbon and energy source. During growth on 1,3-dichlorobenzene, almost stoichiometric amounts of chloride
Heat capacities and densities of some liquid chloro-, bromo-, and bromochloro-substituted benzenes.
Goralski P and Piekarski H.
Journal of Chemical and Engineering Data, 52(2), 655-659 (2007)
J Messner et al.
Chemical communications (Cambridge, England), 50(79), 11705-11708 (2014-08-22)
The isomerisation reaction of 1,4-dichlorobenzene leading to the thermodynamically favoured and technically desired 1,3-dichlorobenzene has been studied comparing highly acidic chloroaluminate melts with organic imidazolium and alkali metal ions. Interestingly, the inorganic melts show much higher reactivity and full recyclability
N Yoshida et al.
Analytical chemistry, 73(18), 4417-4421 (2001-09-29)
A distributed-feedback dye laser has been developed for achieving the efficient multiphoton ionization of chlorobenzene and dichlorobenzene, that is, precursor molecules of dioxins. This tunable picosecond laser with a narrow spectral line width, that is, a nearly transform-limited pulse, provides
T Umemura et al.
Toxicology and applied pharmacology, 137(2), 268-274 (1996-04-01)
The acute hepatotoxicity of isomers of dichlorobenzene (o-, m-, and p-DCB) was compared in livers of male B6C3F1 mice at different time points after a single intragastric administration. The highest doses of o-, m-, and p-DCB administered, 300, 300, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service