Skip to Content
Merck
All Photos(1)

Key Documents

343714

Sigma-Aldrich

Dichlorotris(1,10-phenanthroline)ruthenium(II) hydrate

98%

Synonym(s):

Tris(1,10-phenanthroline)ruthenium(II) chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H24Cl2N6Ru · xH2O
CAS Number:
Molecular Weight:
712.59 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

reaction suitability

core: ruthenium
reaction type: Photocatalysis
reagent type: catalyst

SMILES string

O.Cl[Ru]Cl.c1cnc2c(c1)ccc3cccnc23.c4cnc5c(c4)ccc6cccnc56.c7cnc8c(c7)ccc9cccnc89

InChI

1S/3C12H8N2.2ClH.H2O.Ru/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;;/h3*1-8H;2*1H;1H2;/q;;;;;;+2/p-2

InChI key

WGNGRGQLNAWTJI-UHFFFAOYSA-L

General description

The oxygen sensitivity and phototoxicity of [Ru(Phen)] in chicken embryo chorioallantoic membrane was investigated.

Application

Fluorescence quenching of dichloro(tris-1,10-phenanthroline) ruthenium (II) hydrate is is triggered by oxygen.1 (Ru(phen)3Cl2 may be used to imprint mesoporous molecular sieves (SBA-15).

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianfeng Zhang et al.
Polymers, 12(4) (2020-04-25)
A cost-effective, simple, and time-saving method to fabricate mono-dispersed periodic microsphere structures on substrates with patterned sites is very meaningful due to their significance on various biological studies. Herein, a simple and facile method to fabricate mono-dispersed microsphere arrays on
Oxygen sensor based on the fluorescence quenching of a ruthenium complex immobilized in a biocompatible poly (ethylene glycol) hydrogel.
O' Neal DP, et al.
Sensors null
Molecular imprinting of mesoporous SBA-15 with chiral ruthenium complexes.
Coutinho D,et al
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association null
Oxygen sensor based on the fluorescence quenching of a ruthenium complex immobilized in a biocompatible poly (ethylene glycol) hydrogel.
O' Neal DP, et al.
Sensors, 728-734 null
Marleen Häring et al.
The Journal of organic chemistry, 83(15), 7928-7938 (2018-05-29)
In this work, we demonstrate that useful C-C bond-forming photoredox catalysis can be performed in air using easily prepared gel networks as reaction media to give similar results as are obtained under inert atmosphere conditions. These reactions are completely inhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service