341452
Aluminum phosphate
reagent grade
Synonym(s):
Aluminum monophosphate, Monoaluminum phosphate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
AlO4P
CAS Number:
Molecular Weight:
121.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
reagent grade
form:
powder
Recommended Products
grade
reagent grade
form
powder
density
2.56 g/mL at 25 °C (lit.)
SMILES string
[Al+3].[O-]P([O-])([O-])=O
InChI
1S/Al.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3
InChI key
ILRRQNADMUWWFW-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
General description
Aluminum phosphate is a thermally stable white crystalline solid suitable for high-temperature applications. Additionally, it is an effective flame retardant and is resistant to acids, which enhances its utility in diverse environments. Its unique properties make it suitable for use in energy storage, catalysis, ceramics, and pharmaceuticals.
Application
Aluminum phosphate can be used:
- As a bifunctional additive to fabricate electrode for Li-S batteries. It acts as a lithium polysulfide capturing agent to reduce and stabilize interfacial resistance during cycling.
- As a protective coating on the electrodes of Li-ion batteries to help mitigate issues related to electrode degradation and instability, thereby improving the long-term performance and cycle life of the battery.
- As a precursor to synthesize flame retardant composites.
Physical properties
Nonstoichiometric compound
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Belma Ljutic et al.
Vaccine, 30(19), 2981-2988 (2012-03-03)
We investigated the immunogenicity, stability and adsorption properties of an experimental pneumococcal vaccine composed of three protein vaccine antigens; Pneumococcal histidine triad protein D, (PhtD), Pneumococcal choline-binding protein A (PcpA) and genetically detoxified pneumolysin D1 (PlyD1) formulated with aluminum salt
Shibo Jiang et al.
Expert review of vaccines, 11(12), 1405-1413 (2012-12-21)
A subunit vaccine, RBD-S, is under development to prevent severe acute respiratory syndrome (SARS) caused by SARS coronavirus (SARS-CoV), which is classified by the US NIH as a category C pathogen. This vaccine is comprised of a recombinant receptor-binding domain
Fikru Tafesse et al.
Talanta, 83(5), 1491-1495 (2011-01-18)
A highly selective and sensitive phosphate sensor has been fabricated by constructing a crystal disk consisting of variable mixtures of aluminium powder (Al), aluminium phosphate (AlPO(4)) and powdered copper (Cu). The membrane sensor exhibits linear potential response in the concentration
Unified internal architecture and surface barriers for molecular diffusion of microporous crystalline aluminophosphates.
Lukasz Karwacki et al.
Angewandte Chemie (International ed. in English), 49(38), 6790-6794 (2010-08-18)
H M El Sahly et al.
Clinical and vaccine immunology : CVI, 17(10), 1552-1559 (2010-08-13)
Erythrocyte binding antigen region II (EBA-175) is a conserved antigen of Plasmodium falciparum that is involved in binding of the parasite to the host's erythrocytes. We evaluated the safety and immunogenicity of a recombinant EBA-175 vaccine with aluminum phosphate adjuvant
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service