Skip to Content
Merck
All Photos(1)

Key Documents

338567

Sigma-Aldrich

Sulfosuccinic acid solution

70 wt. % in H2O

Synonym(s):

2-Sulfobutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(SO3H)COOH
CAS Number:
Molecular Weight:
198.15
Beilstein:
1727687
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

concentration

70 wt. % in H2O

refractive index

n20/D 1.449

density

1.438 g/mL at 25 °C

SMILES string

OC(=O)CC(C(O)=O)S(O)(=O)=O

InChI

1S/C4H6O7S/c5-3(6)1-2(4(7)8)12(9,10)11/h2H,1H2,(H,5,6)(H,7,8)(H,9,10,11)

InChI key

ULUAUXLGCMPNKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Modification of nanocellulose membrane by impregnation method with sulfosuccinic acid for direct methanol fuel cell applications: This study examines the use of sulfosuccinic acid in various concentrations to improve the properties of nanocellulose membranes for fuel cells (Sriruangrungkamol & Chonkaew, 2021).
  • A high-strength polyvinyl alcohol hydrogel membrane crosslinked by sulfosuccinic acid for strontium removal via filtration: Research focusing on using sulfosuccinic acid to enhance polyvinyl alcohol hydrogel membranes for effective strontium filtration (Yoon et al., 2019).
  • Cross-linked poly (vinyl alcohol)/sulfosuccinic acid polymer as an electrolyte/electrode material for H2–O2 proton exchange membrane fuel cells: This paper discusses the application of sulfosuccinic acid in creating cross-linked polyvinyl alcohol for fuel cell membranes (Ebenezer et al., 2016).
  • Preparation of chitosan/polyvinyl alcohol blended films containing sulfosuccinic acid as the crosslinking agent using UV curing process: An investigation into using sulfosuccinic acid to crosslink chitosan/polyvinyl alcohol films for improved mechanical properties (Yun et al., 2017).
  • Investigation on resistive humidity sensing in sulfosuccinic acid doped polyaniline films: This research explores the potential of sulfosuccinic acid doped polyaniline films for humidity sensing applications (Biswas et al., 2021).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Kim et al.
Water research, 160, 445-453 (2019-06-08)
Technologies capable of selective removal of target contaminants from water are highly desirable to achieve "fit-for-purpose" treatment. In this study, we developed a simple yet highly effective method to achieve calcium-selective removal in an electrosorption process by coating the cathode
T P Alekseeva et al.
Izvestiia Akademii nauk. Seriia biologicheskaia, (4)(4), 403-410 (2010-08-31)
Comparative study of chitosan wound healing properties and its synthesized derivatives in MC-100 gel was carried out using the model of experimental full thickness skin wounds. It was determined that N-sulfosuccinoyl chitosan derivatives added into the gel in a concentration
Jeffrey Childs et al.
Journal of contaminant hydrology, 82(1-2), 1-22 (2005-10-20)
This study reports on a surfactant-based flood for tetrachloroethylene (PCE) removal from a control test cell at the Dover National Test Site. The surfactant formulation (sodium dihexyl sulfosuccinate (Aerosol-MA or AMA), isopropanol and calcium chloride) was able to achieve a
Permanent enzyme microencapsulation in reverse micellar media.
L Robledo et al.
Annals of the New York Academy of Sciences, 750, 89-93 (1995-03-31)
B A Simmons et al.
Journal of the American Chemical Society, 123(10), 2414-2421 (2001-07-18)
Dry reverse micelles of the anionic twin-tailed surfactant bis(2-ethylhexyl) sulfosuccinate (AOT) dissolved in nonpolar solvents spontaneously form an organogel when p-chlorophenol is added in a 1:1 AOT:phenol molar ratio. The solvents used were benzene, toluene, m-xylene, 2,2,4-trimethylpentane (isooctane), decane, dodecane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service