Skip to Content
Merck
All Photos(3)

Documents

338370

Sigma-Aldrich

Pentamethylcyclopentadienylrhodium(III) chloride dimer

97%

Synonym(s):

1,3-Cyclopentadiene, 1,2,3,4,5-pentamethyl-, rhodium complex, Bis(dichloro(pentamethylcyclopentadienyl)rhodium), Bis(pentamethylcyclopentadienylrhodium dichloride), Dichloro(pentamethylcyclopentadienyl)rhodium(II) dimer, Tetrachlorobis(pentamethylcyclopentadienyl)dirhodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Rh(C5Me5)Cl2]2
CAS Number:
Molecular Weight:
618.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: rhodium
reaction type: C-H Activation
reagent type: catalyst
reaction type: C-Mannosylation

SMILES string

Cl[Rh]Cl.Cl[Rh]Cl.C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.4ClH.2Rh/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*1-5H3;4*1H;;/q;;;;;;2*+2/p-4

InChI key

QNIVKTTWBMFSBR-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Umicore Precatalysts for Asymmetric and Cross-Coupling Catalysis

Catalyst for:
  • Oxidative olefination reactions
  • C-C bond cleavage of secondary alcohols
  • Ortho C-H olefination of phenol derivatives
  • Oxidative annulation of pyridines
  • Oxidative ortho-acylation of benzamides with aldehydes via direct functionalization of the sp2 C-H bond
  • Amidation reactions
  • Reductive alkylation reactions
Pentamethylcyclopentadienylrhodium(III) chloride dimer can be used as a catalyst for:
  • Oxidative olefination reactions
  • C-C bond cleavage of secondary alcohols
  • Ortho C-H olefination of phenol derivatives
  • Oxidative annulation of pyridines
  • Oxidative ortho-acylation of benzamides with aldehydes via direct functionalization of the sp2 C-H bond
  • Amidation reactions
  • Reductive alkylation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentamethylcyclopentadienylrhodium(III) chloride dimer.
Too P C and Chiba S
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2013)
(?5?Pentamethylcyclopentadienyl)Rhodium and ?Iridium Compounds.
White C, et al.
Inorganic Syntheses, 29, 228-234 (2007)
Pentamethylcyclopentadienylrhodium(III) chloride dimer.
Too P C and Chiba S
e-EROS Encyclopedia of Reagents for Organic Synthesis (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service