Skip to Content
Merck
All Photos(1)

Documents

334103

Sigma-Aldrich

Silicon nitride

powder, ≥99.9% trace metals basis

Synonym(s):

Trisilicon tetranitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.9% trace metals basis

form

powder

particle size

<1 μm

density

3.44 g/mL at 25 °C (lit.)

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Silicon nitride based ceramics possess high fracture toughness, hardness and wear resistance, resistance to corrosion, thermal stability. Two known polymorphs of silicon nitride exist, alpha and beta Si3N4, both have hexagonal structure. A study shows that a third state, cubic spinel structure, has also been synthesized.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of cubic silicon nitride
Zerr A, et al
Nature, 400, 340-342 null
Silicon Nitride and Related Materials
Riley FR
Journal of the American Ceramic Society. American Ceramic Society, 83(2), 245-265 (2000)
Chang-Hyun Park et al.
Optics express, 20(21), 23769-23777 (2012-11-29)
A nanophotonic polarization-independent visible wavelength filter is presented, incorporating a symmetric metal-dielectric resonant structure on quartz substrate, where a sub-wavelength grating, made up of a two-dimensional array of Al square sheets, is integrated with a Si(3)N(4) slab waveguide via an
Simone P Rodrigues et al.
Journal of materials science. Materials in medicine, 24(1), 231-239 (2012-10-12)
Commercial femoral head prostheses (cobalt-chromium alloy, yttria partially stabilized zirconia (Y-PSZ) and alumina) and new silicon nitride ceramic ones (nanocrystalline diamond coated and uncoated) were compared in terms of artifact level production by computed tomography (CT). Pelvis examination by CT
Madeline J Dukes et al.
Chemical communications (Cambridge, England), 49(29), 3007-3009 (2013-03-06)
Gold nanorods are widely known for their photothermal properties to treat solid tumors. Our work demonstrates the unrealized capacity to image these reagents in liquid at high resolution using Transmission Electron Microscopy (TEM). Here we perform the first atomic measurements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service