Skip to Content
Merck
All Photos(2)

Key Documents

330647

Sigma-Aldrich

2,2′-Bis(bromomethyl)-1,1′-biphenyl

99%

Synonym(s):

1-(Bromomethyl)-2-[2-(bromomethyl)phenyl]benzene, 2,2′-Bis(bromomethyl)-1,1′-biphenyl, 2,2′-Bis(bromomethyl)biphenyl, 2,2′-Di(bromomethyl)biphenyl, o,o′-Bis(bromomethyl)biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4C6H4CH2Br
CAS Number:
Molecular Weight:
340.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

90-92 °C (lit.)

functional group

bromo

SMILES string

BrCc1ccccc1-c2ccccc2CBr

InChI

1S/C14H12Br2/c15-9-11-5-1-3-7-13(11)14-8-4-2-6-12(14)10-16/h1-8H,9-10H2

InChI key

VDXGURZGBXUKES-UHFFFAOYSA-N

Application

2,2′-Bis(bromomethyl)-1,1′-biphenyl was used in the preparation of mononuclear rhodium(III) complex with a nine-membered S, S-chelate ring.[1] It was also used in the preparation of 1,1′-binaphthyl-substituted α-aminoisobutyric acid, a new chiral atropoisomeric α,α-disubstituted glycine.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuto Yoshinari et al.
Inorganic chemistry, 47(17), 7450-7452 (2008-08-07)
Treatment of fac-[Rh(aet) 3] (aet = 2-aminoethanethiolate) with 2,2'-bis(bromomethyl)-1,1'-biphenyl gave a mononuclear rhodium(III) complex with a nine-membered S, S-chelate ring, fac-[Rh(aet)(L)] (2+) ([ 1] (2+), L = 2,2'-bis(2-aminoethylthiomethyl)-1,1'-biphenyl). Complex [ 1] (2+) afforded a pair of atrop diastereomers, Delta SS(
Novel α, α-disubstituted a-aminoacids with axial dissymmetry and their N-or C-protected derivatives.
Mazaleyrat J-P, et al.
Tetrahedron Asymmetry, 8(4), 619-631 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service