328669
Tetravinyltin
97%
Synonym(s):
Stannane, Tetraethenylstannane, Tetrakis(ethenyl)stannane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Sn(CH=CH2)4
CAS Number:
Molecular Weight:
226.89
Beilstein:
3535492
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
form
liquid
reaction suitability
core: tin
refractive index
n20/D 1.501 (lit.)
bp
160-163 °C (lit.)
density
1.246 g/mL at 25 °C (lit.)
SMILES string
C=C[Sn](C=C)(C=C)C=C
InChI
1S/4C2H3.Sn/c4*1-2;/h4*1H,2H2;
InChI key
MZIYQMVHASXABC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service