326259
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12INO2
CAS Number:
Molecular Weight:
293.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
114-116 °C (lit.)
functional group
ester
SMILES string
[I-].CC[n+]1ccc(cc1)C(=O)OC
InChI
1S/C9H12NO2.HI/c1-3-10-6-4-8(5-7-10)9(11)12-2;/h4-7H,3H2,1-2H3;1H/q+1;/p-1
InChI key
NGEAJXXGUZQCPN-UHFFFAOYSA-M
General description
Ion association/dissociation behavior of 1-ethyl-4-(methoxycarbonyl)pyridinium iodide in a number of ionic and molecular liquids by mathematical and computational modelling was reported.
Application
1-Ethyl-4-(methoxycarbonyl)pyridinium iodide was employed as probe for micropolarity determinations. It was also used in preparation of 1-ethyl-4-(methoxycarbonyl)pyridinium tetrakis(4-phenoxyphenyl)borate.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Terhi Alaviuhkola et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(7), 2071-2080 (2005-01-28)
Five aromatic borate anions, namely tetrakis(4-phenoxyphenyl)borate (1), tetrakis(biphenyl)borate (2), tetrakis(2-naphthyl)borate (3), tetrakis(4-phenylphenol)borate (4), and tetrakis(4-phenoxy)borate (5), have been prepared and tested as ion-recognition sites in chemical sensors for certain aromatic cations and metal ions. To gain further insight into the
Salts dissolved in salts: ionic liquid mixtures.
Lui MY, et al.
Chemical Science, 2(8), 1491-1496 (2011)
Micropolarity of sodium bis (2-ethylhexyl) sulfosuccinate reverse micelles prepared in supercritical ethane and near-critical propane.
Shervani Z and Ikushima Y.
Coll. Polymer Sci., 277(6), 595-600 (1999)
Sophia Jiyuan Zhang et al.
Applied and environmental microbiology, 85(6) (2019-02-03)
A cup of coffee is the final product of a complex chain of operations. Wet postharvest processing of coffee is one of these operations, which involves a fermentation that inevitably has to be performed on-farm. During wet coffee processing, the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service